Skip to Content
Merck
All Photos(3)

Key Documents

901102

Sigma-Aldrich

PPT

sublimed

Synonym(s):

2,8-Bis(diphenylphosphoryl)dibenzo[b,d]thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H26O2P2S
CAS Number:
Molecular Weight:
584.60
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

Quality Level

form

solid

color

white

SMILES string

O=P(C1=CC=CC=C1)(C2=CC=CC=C2)C(C=C3)=CC4=C3SC5=C4C=C(P(C6=CC=CC=C6)(C7=CC=CC=C7)=O)C=C5

InChI

1S/C36H26O2P2S/c37-39(27-13-5-1-6-14-27,28-15-7-2-8-16-28)31-21-23-35-33(25-31)34-26-32(22-24-36(34)41-35)40(38,29-17-9-3-10-18-29)30-19-11-4-12-20-30/h1-26H

InChI key

ZCJJIQHVZCFSGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A New Molecular Design Based on Thermally Activated Delayed Fluorescence for Highly Efficient Organic Light Emitting Diodes.
Rajamalli P, et al.
Journal of the American Chemical Society, 138(2), 628-634 (2016)
Thermally activated delayed fluorescence emitters with a m,m-di-tert-butyl-carbazolyl benzoylpyridine core achieving extremely high blue electroluminescence efficiencies.
Rajamalli P, et al.
Journal of Material Chemistry C, 5(11), 2919-2926 (2017)
Ryuhei Furue et al.
Angewandte Chemie (International ed. in English), 55(25), 7171-7175 (2016-05-05)
Luminescent materials consisting of boron clusters, such as carboranes, have attracted immense interest in recent years. In this study, luminescent organic-inorganic conjugated systems based on o-carboranes directly bonded to electron-donating and electron-accepting π-conjugated units were elaborated as novel optoelectronic materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service