SCM123
ChondroMAX Differentiation Medium
Synonym(s):
MSC chondrocyte media, chondrocyte differentiation media, chondrogenesis media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
shipped in
dry ice
storage temp.
−20°C
General description
Mesenchymal stem cells (MSCs) are adult stem cells, which have the capacity for multi-lineage differentiation, giving rise to a variety of mesenchymal phenotypes such as osteoblasts (bone), adipocytes (fat), and chondrocytes (cartilage). Articular hyaline cartilage has poor regenerative capacity, and the loss of its function is, in the long term, often painful and debilitating. Therefore, attempts have been made to study chondrogenesis or to replace damaged cartilage tissue with MSC derived stem cells.
ChondroMAX Differentiation Medium is a ready-to-use xeno-free mesenchymal stem cell chondrogenesis differentiation media. ChrondroMAX produces cells with elevated levels of sulfated proteoglycans, analyzed by Alcian-Blue staining, after 3 weeks of MSC pellet differentiation compared to other chrondrogenesis differentiation media. ChondroMAX is an excellent research tool to study chondrocyte differentiation, chondrogenesis and wound healing.
ChondroMAX Differentiation Medium is a ready-to-use xeno-free mesenchymal stem cell chondrogenesis differentiation media. ChrondroMAX produces cells with elevated levels of sulfated proteoglycans, analyzed by Alcian-Blue staining, after 3 weeks of MSC pellet differentiation compared to other chrondrogenesis differentiation media. ChondroMAX is an excellent research tool to study chondrocyte differentiation, chondrogenesis and wound healing.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service