TMS-009
Safranin-O Staining Solution
Safranin-O is often used to identify chondrocytes that have been derived from both human and rodent mesenchymal stem cells (MSCs).
Synonym(s):
Basic Red 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
Quality Level
concentration
0.1 % (w/v) in distilled water
technique(s)
cell based assay: suitable
cell culture | stem cell: suitable
cell differentiation: suitable
General description
Safranin-O, also known as basic red 2, is a biological stain used in histology and cytology. Safranin is used as a counterstain in some staining protocols, colouring all cell nuclei red. It can also be used for the detection of cartilage, mucin and mast cell granules. Safranin-O is often used to identify chondrocytes that have been derived from both human and rodent mesenchymal stem cells.
Application
Research Category
Stem Cell Research
Stem Cell Research
Research Sub Category
Osteobiology
Arthritis
Osteobiology
Arthritis
Quality
Solution is red with no particulates present. Solution is suitable for endpoint cell culture assays.
Physical form
Product is presented as a 0.1% w/v solution diluted in distilled water.
Storage and Stability
Store at room temperature for up to 4 months from date of receipt. If Safraninin-O solution forms a precipitate, remove particulates by passage through a 0.22 or 0.45-micron filter.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service