SCM045
Human Mesenchymal-XF Expansion Medium
Xeno-free mesenchymal stem cell media specially formulated for the rapid expansion of human mesenchymal stem cells isolated from a variety of sources, including adipose and bone marrow tissues.
Synonym(s):
Human MSC Culture Media
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Chemicon®
technique(s)
cell culture | stem cell: suitable
input
sample type mesenchymal stem cell(s)
shipped in
dry ice
General description
The Human Mesenchymal-XF Expansion Medium is a xeno-free medium specially formulated for the rapid expansion of human mesenchymal stem cells isolated from a variety of sources, including adipose and bone marrow tissues. The medium contains 2% human serum, is free of animal-derived components and supports the growth of human MSCs at rates equal to or greater than other commercial xeno-free/serum-free media and media supplemented with 2-10% FBS. Human MSCs cultured in the medium retain self-renewal and multi-lineage differenitation potential. The Human Mesenchymal-XF Expansion Medium does not contain any antimicrobials, phenol red, or any other components that can cause cell stress and “masking effects” that may influence experimental results.
Application
A separate ECM solution is not required to facilitate cell attachment using the human mesenchymal-XF expansion medium.
Research Category
Stem Cell Research
Stem Cell Research
Research Sub Category
Mesenchymal Stem Cells
Mesenchymal Stem Cells
Components
All components used within the media are of human origin and do not contain any animal derived components.
Quality
These products are manufactured with the highest quality of raw materials, with exacting standards and production procedures to ensure lot-to-lot consistency. Every lot of medium is extensively tested for the following parameters:
Sterility Testing: Negative for bacterial and fungal growth
pH: 7.5 ± 0.3
Cell Testing: Rate of proliferation and morphology
Osmolality: 255 ± 15 mOsm
Endotoxin: <0.5 EU/mL
Sterility Testing: Negative for bacterial and fungal growth
pH: 7.5 ± 0.3
Cell Testing: Rate of proliferation and morphology
Osmolality: 255 ± 15 mOsm
Endotoxin: <0.5 EU/mL
Physical form
The Human Mesenchymal-XF Expansion Medium is a complete medium and contains all the necessary growth factors and supplements to support human mesenchymal stem cell growth.
Storage and Stability
Frozen medium should be stored at -20ºC until expiration date.
Thawed medium can be stored at 2 to 8ºC for up to 1 month.
Multiple freeze/thaw cycles are not recommended
Thawed medium can be stored at 2 to 8ºC for up to 1 month.
Multiple freeze/thaw cycles are not recommended
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service