Skip to Content
Merck
All Photos(1)

Key Documents

S101

Sigma-Aldrich

(R)-(+)-SKF-38393 hydrochloride

≥98% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO2 · HCl
CAS Number:
Molecular Weight:
291.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

optical activity

[α]22/D +16.1°, c = 1.2 in methanol(lit.)

storage condition

desiccated

color

off-white to light tan

solubility

0.1 M HCl: 1.2 mg/mL
ethanol: 3.4 mg/mL
H2O: 5 mg/mL
aqueous base: unstable

SMILES string

Cl.Oc1cc2CCNC[C@H](c3ccccc3)c2cc1O

InChI

1S/C16H17NO2.ClH/c18-15-8-12-6-7-17-10-14(13(12)9-16(15)19)11-4-2-1-3-5-11;/h1-5,8-9,14,17-19H,6-7,10H2;1H/t14-;/m1./s1

InChI key

YEWHJCLOUYPAOH-PFEQFJNWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(+)-SKF-38393 hydrochloride has been used as a D1 dopamine receptor agonist to study its effect on the sleep-wake pattern of macaques rendered parkinsonian with 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP).

Biochem/physiol Actions

(R)-(+)-SKF-38393 hydrochloride is a D1 dopamine receptor agonist and an active enantiomer of (±)-SKF-38393. It enhances pertussis toxin-insensitive and protein kinase A-mediated glutamate release in the hippocampal neurons. SKF-38393, a benzazepine derivative, exhibits anorectic effects.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Light sensitive

Reconstitution

Solutions may be stored for several days at 4 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carole Hyacinthe et al.
Neurobiology of disease, 63, 20-24 (2013-11-12)
Both excessive daytime sleepiness (EDS) and rapid eye movement (REM) sleep deregulation are part of Parkinson's disease (PD) non-motor symptoms and may complicate dopamine replacement therapy. We report here that dopamine agonists act differentially on sleep architecture in the 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine
A Bouron et al.
Neuroscience, 94(4), 1063-1070 (2000-01-07)
The present study was undertaken to better assess the role of dopamine on exocytosis. Since direct activation of adenylate cyclase (e.g., with forskolin) enhances neurotransmitter release it was of interest to see whether the activation of D1-type dopamine receptors, which
M R Zarrindast et al.
Pharmacology, 87(1-2), 85-89 (2011-01-19)
Both the dopamine receptor D(1) agonist SKF 38393 and the antagonist SCH 23390 are benzazepine derivatives that have been widely used as pharmacological tools and radioligands. Evidence suggests that behavioral effects of both compounds do not always correspond to their
E Ongini et al.
Life sciences, 37(24), 2327-2333 (1985-12-16)
The dopamine D-1 receptor agonist SKF 38393 dose-dependently (2.5-10 mg/kg) induced desynchronization of the electroencephalographic (EEG) activity and behavioral arousal in both rabbits and rats. Unlike apomorphine, SKF 38393 elicited no signs of stereotyped behavior in rabbits and minimal effects
C Kaiser et al.
Journal of medicinal chemistry, 25(6), 697-703 (1982-06-01)
Resolution of the unique dopamine receptor agonist 2,3,4,5-tetrahydro-7,8-dihydroxy-1-phenyl-1H-3-benzazepine (1) was achieved by a stereospecific multistep conversion of the readily separated enantiomers of its O,O,N-trimethylated precursor 2. The absolute stereochemistry of the antipodes of 2-MeI was determined by single-crystal X-ray diffractometric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service