Skip to Content
Merck
All Photos(2)

Key Documents

C130

Sigma-Aldrich

Chloro-APB hydrobromide

solid

Synonym(s):

(±)-6-Chloro-7,8-dihydroxy-3-allyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrobromide, (±)-SKF-82958 hydrobromide, 6-Chloro-N-allyl-SKF-38393 hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20ClNO2 · HBr
CAS Number:
Molecular Weight:
410.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

tan

solubility

H2O: 3.7 mg/mL
aqueous base: unstable
ethanol: soluble

storage temp.

2-8°C

SMILES string

Br.Oc1cc2C(CN(CCc2c(Cl)c1O)CC=C)c3ccccc3

InChI

1S/C19H20ClNO2.BrH/c1-2-9-21-10-8-14-15(11-17(22)19(23)18(14)20)16(12-21)13-6-4-3-5-7-13;/h2-7,11,16,22-23H,1,8-10,12H2;1H

InChI key

WLXGFAVTAAQOFH-UHFFFAOYSA-N

Gene Information

human ... DRD1(1812)

Related Categories

Application

Chloro-APB hydrobromide has been used to pharmacologically stimulate D1 dopamine receptors and study its influence on locomotor response in fat mass and obesity-associated protein (FTO) deficient dopamine D1 receptor-expressing medium spiny neurons (D1 MSNs) of mice.

Biochem/physiol Actions

Chloro-APB hydrobromide, also known as SKF 82958, is a D1 dopamine receptor agonist. D1 receptor agonists prevent and reverse fentanyl-induced respiratory depression, without impairing opioid analgesia, in cats.

Caution

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Liad Mann et al.
The international journal of neuropsychopharmacology, 11(4), 533-539 (2008-01-22)
Lithium ions' inhibition of adenylyl cyclase (AC) has not been previously studied for the newly discovered AC isoforms. COS7 cells were transfected with each of the nine membrane-bound AC isoforms cDNAs with or without D1- or D2-dopamine receptor cDNA. AC
Neil E Paterson et al.
European journal of pharmacology, 616(1-3), 73-80 (2009-06-06)
Harmaline-induced tremor in rodents is a model of essential tremor. We utilized a novel assay to quantify tremor activity in mice and found that tremor activity was dependent on harmaline dose. The first-line clinical essential tremor treatments propranolol, primidone and
Stefanie Dedeurwaerdere et al.
The Journal of pharmacology and experimental therapeutics, 339(1), 210-217 (2011-07-12)
Phosphodiesterase 10A (PDE10A) inhibitors have recently been proposed as a new therapy for schizophrenia. The aim of this study was to enhance our understanding of the role of PDE10A inhibitors and potentially identify a clinically useful mechanistic/functional biomarker by using
Peter M Lalley
American journal of physiology. Regulatory, integrative and comparative physiology, 289(1), R45-R51 (2005-02-12)
Opioids depress respiration and decrease chest wall compliance. A previous study in this laboratory showed that dopamine-D(1) receptor (D(1)R) agonists restored phrenic nerve activity after arrest by fentanyl in immobilized, mechanically ventilated cats. The reinstated phrenic nerve rhythm was slower
Kenneth D Carr et al.
Psychopharmacology, 202(4), 731-743 (2008-10-09)
Previous studies have suggested that chronic food restriction (FR) increases sensitivity of a neural substrate for drug reward. The neuroanatomical site(s) of key neuroadaptations may include nucleus accumbens (NAc) where changes in D-1 dopamine (DA) receptor-mediated cell signaling and gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service