Skip to Content
Merck
All Photos(4)

Key Documents

D047

Sigma-Aldrich

(±)-SKF-38393 hydrochloride

crystalline, ≥98% (HPLC)

Synonym(s):

(±)-1-Phenyl-2,3,4,5-tetrahydro-(1H)-3-benzazepine-7,8-diol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO2 · HCl
CAS Number:
Molecular Weight:
291.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

crystalline

storage condition

desiccated
protect from light

color

white

solubility

H2O: >5 mg/mL
aqueous base: soluble (unstable)

SMILES string

Cl.Oc1cc2CCNCC(c3ccccc3)c2cc1O

InChI

1S/C16H17NO2.ClH/c18-15-8-12-6-7-17-10-14(13(12)9-16(15)19)11-4-2-1-3-5-11;/h1-5,8-9,14,17-19H,6-7,10H2;1H

InChI key

YEWHJCLOUYPAOH-UHFFFAOYSA-N

Gene Information

human ... DRD1(1812)

Looking for similar products? Visit Product Comparison Guide

Application

(±)-SKF-38393 hydrochloride has been used as a selective D1 receptor agonist as a part of dopaminergic treatment.[1][2] It has also been used to study its effect on prolactin in male baboons.[3]

Biochem/physiol Actions

(±)-SKF-38393 is a D1 dopamine receptor agonist. It is known to promote the production of glutamate in the hippocampus.[4]

Caution

This material is hygroscopic. Air-sensitive. Light-sensitive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The D1 dopamine receptor agonist SKF-38393 stimulates the release of glutamate in the hippocampus
Bouron A and Reuter H
Neuroscience, 94(4), 1063-1070 (1999)
Prolactin and fMRI response to SKF38393 in the baboon
Miller B, et al.
PeerJ, 1(5), e195-e195 (2013)
Stimulation of dopamine D1 receptor improves learning capacity in cooperating cleaner fish
Messias JPM, et al.
Proc. Royal Soc. Lond. B., 283(1823), 20152272-20152272 (2016)
David Kabelik et al.
Neuropharmacology, 58(1), 117-125 (2009-06-23)
Recent experiments demonstrate that aggressive competition for potential mates involves different neural mechanisms than does territorial, resident-intruder aggression. However, despite the obvious importance of mate competition aggression, we know little about its regulation. Immediate early gene experiments show that in
Dopamine disruption increases cleanerfish cooperative investment in novel client partners
Soares MC, et al.
Royal Society open science, 4(5), 160609-160609 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service