Skip to Content
Merck
All Photos(2)

Documents

S0758

Sigma-Aldrich

Sulfaphenazole

≥98%

Synonym(s):

4-Amino-N-(1-phenyl-1H-pyrazol-5-yl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N4O2S
CAS Number:
Molecular Weight:
314.36
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

powder

solubility

ethanol: 25 mg/mL, clear, colorless to yellow

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2ccnn2-c3ccccc3

InChI

1S/C15H14N4O2S/c16-12-6-8-14(9-7-12)22(20,21)18-15-10-11-17-19(15)13-4-2-1-3-5-13/h1-11,18H,16H2

InChI key

QWCJHSGMANYXCW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Sulfaphenazole has been used as a positive control to inhibit cytochrome P450 2C9 (cyp2c9) to quantify Rhodiola rosea inhibition. It has also been used as cytochrome P450 2C9 (cyp2c9) inhibitor in endothelial cells and microsomal preparations.

Biochem/physiol Actions

Antibacterial. Specific inhibitor of CYP2C9. Blocks pro-inflammatory and atherogenic effects of linoleic acid (increase in oxidative stress and activation of AP-1) mediated by CYP2C9. Specific inhibitor of CYP2C9. Blocks pro-inflammatory and atherogenic effects of linoleic acid (increase in oxidative stress and activation of AP-1) mediated by CYP2C9. Inhibits bradykinin-induced tPA release.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accelerating Drug Discovery Efforts for Trypanosomatidic Infections Using an Integrated Transnational Academic Drug Discovery Platform
Moraes CB, et al.
SLAS discovery, 24(3), 346-361 (2019)
Ralf Heim et al.
Journal of medicinal chemistry, 51(16), 5064-5074 (2008-08-05)
Recently, we reported on the development of potent and selective inhibitors of aldosterone synthase (CYP11B2) for the treatment of congestive heart failure and myocardial fibrosis. A major drawback of these nonsteroidal compounds was a strong inhibition of the hepatic drug-metabolizing
Zhiping Jin et al.
Basic & clinical pharmacology & toxicology, 130(5), 592-605 (2022-03-16)
Anlotinib is a small molecule of novel tyrosine kinase inhibitor initially approved to treat non-small cell lung cancer in China. Drug-drug interaction (DDI) is an extrinsic factor important for the appropriate use of anlotinib in clinical practice. In vitro experiments
Altered purinergic signaling in uridine adenosine tetraphosphate-induced coronary relaxation in swine with metabolic derangement
Zhou Z, et al.
Purinergic Signaling, 13(3), 319-329 (2017)
Vienna E Brunt et al.
The Journal of physiology, 590(15), 3523-3534 (2012-06-08)
While it is accepted that NO is responsible for ∼60% of the plateau in cutaneous thermal hyperaemia, a large portion of the response remains unknown. We sought to determine whether the remaining ∼40% could be attributed to EDHF-mediated activation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service