Skip to Content
Merck
All Photos(3)

Key Documents

UC175

Sigma-Aldrich

(S)-(+)-Mephenytoin

≥98% (HPLC), solid, CYP2B6 & CYP2C19 substrate

Synonym(s):

(S)-(+)-5-Ethyl-3-methyl-5-phenyl-2,4-imidazolidinedione, (S)-(+)-5-Ethyl-3-methyl-5-phenylhydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Product Name

(S)-(+)-Mephenytoin, solid, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

solid

color

off-white

mp

135-138 °C

solubility

DMSO: soluble

storage temp.

2-8°C

SMILES string

CC[C@]1(NC(=O)N(C)C1=O)c2ccccc2

InChI

1S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8H,3H2,1-2H3,(H,13,16)/t12-/m0/s1

InChI key

GMHKMTDVRCWUDX-LBPRGKRZSA-N

Application

S-Mephenytoin has been used as a CYP2C19 substrate for the analysis of cytochrome P450 metabolism. S-Mephenytoin has also been used as probe substrates for LC/MS-based analysis of relative activity factor (RAF).

Biochem/physiol Actions

CYP2B6 & CYP2C19 substrate. Methenytoin isomer. Anticonvulsant; antiepileptic.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

(S)-(+)-Mephenytoin is soluble in DMSO.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Brandon S Pybus et al.
Malaria journal, 11, 259-259 (2012-08-04)
The 8-aminoquinoline (8AQ) drug primaquine (PQ) is currently the only approved drug effective against the persistent liver stage of the hypnozoite forming strains Plasmodium vivax and Plasmodium ovale as well as Stage V gametocytes of Plasmodium falciparum. To date, several
Stefan J Dekker et al.
British journal of pharmacology, 176(3), 466-477 (2018-11-18)
The aim of this study was to characterize the human cytochrome P450s (CYPs) involved in oxidative bioactivation of flucloxacillin to 5-hydroxymethyl flucloxacillin, a metabolite with high cytotoxicity towards biliary epithelial cells. The CYPs involved in hydroxylation of flucloxacillin were characterized
Bent H Hellum et al.
Basic & clinical pharmacology & toxicology, 105(1), 58-63 (2009-04-18)
The aim of this study was to evaluate in vitro the dose-dependent induction potential of six commonly used trade herbal products on CYP2C19 and CYP2E1 metabolic activities in cultured human hepatocytes. S-mephenytoin and chlorzoxazone were used as specific CYP substrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service