Skip to Content
Merck
All Photos(1)

Key Documents

22600

Sigma-Aldrich

Quinidine

crystallized, ≥98.0% (dried material, NT)

Synonym(s):

β-Quinidine, (+)-Quinidine, (9S)-6′-Methoxycinchonan-9-ol, Chinidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2
CAS Number:
Molecular Weight:
324.42
Beilstein:
91866
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (dried material, NT)

form

solid

optical activity

[α]20/D +265±5°, c = 0.8% in ethanol (dry matter)

quality

crystallized

impurities

≤2% water
~10% hydroquinidine (HPLC)

mp

168-172 °C (lit.)

functional group

hydroxyl

SMILES string

COc1ccc2nccc([C@H](O)C3CC4CCN3C[C@@H]4C=C)c2c1

InChI

1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19+,20-/m0/s1

InChI key

LOUPRKONTZGTKE-LHHVKLHASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinidine is an isomer of quinine.

Application

Quinidine may be used in the preparation of O-trimethylsilylquinidine by reacting trimethylsilyl chloride.

Biochem/physiol Actions

Class IA antiarrhythmic; potassium channel blocker.

Other Notes

Chiral catalyst, used e.g. in highly enantioselective [2+2] cycloadditions

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 50, 1977-1977 (1985)
P.E. Ketelaar et al.
Tetrahedron Letters, 26, 4665-4665 (1985)
Organocatalytic enantioselective synthesis of bicyclic ?-lactones from aldehyde acids via nucleophile-catalyzed aldol-lactonization (NCAL).
Nguygen et al.
Organic Syntheses, 88, 121-137 (2011)
Sami Viskin et al.
Journal of the American College of Cardiology, 61(23), 2383-2387 (2013-04-16)
The aim of this study was to determine the availability of quinidine throughout the world. Quinidine is the only oral medication that is effective for preventing life-threatening ventricular arrhythmias due to Brugada syndrome and idiopathic ventricular fibrillation. However, because of
Bernard Belhassen et al.
Circulation, 110(13), 1731-1737 (2004-09-24)
Automatic implantable cardioverter-defibrillator therapy is considered the only effective treatment for high-risk patients with Brugada syndrome. Quinidine depresses I(to) current, which may play an important role in the arrhythmogenesis of this disease. The effects of quinidine bisulfate (mean dose, 1483+/-240

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service