Skip to Content
Merck
All Photos(1)

Key Documents

P3619

Sigma-Aldrich

Phosphate Buffer Solution

1.0 M, pH 7.4 (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
eCl@ss:
32129211
NACRES:
NA.25
Pricing and availability is not currently available.

form

liquid

Quality Level

concentration

1.0 M

pH

7.4 (25 °C)

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

Application

Phosphate Buffer Solution has been used:
  • to study sperm motility in different culture media[1]
  • for protein extraction[2]
  • as a reagent in blocking buffer for immunolabeling[3]

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Characterization and cryopreservation of Amur leopard cats (Prionailurus bengalensis euptilurus) semen collected by urethral catheterization
Jeong DH, et al.
Theriogenology, 119, 91-95 (2018)
Immunogold Electron Microscopy of the Autophagosome Marker LC3
Mattoscio D, et al.
PLoS Pathogens (2017)
Dong-Hyuk Jeong et al.
Theriogenology, 119, 91-95 (2018-07-10)
The Amur leopard cat (Prionailurus bengalensis euptilurus) is a globally endangered species, and there is thus an urgent need to increase its population. The objectives of this study were to: (1) confirm the utility of urethral catheterization (UC) for semen
G Lê-Bury et al.
Biology of the cell, 112(3), 92-101 (2020-01-11)
Non-typhoidal Salmonella (NTS) causes a severe invasive syndrome (iNTS disease) described in HIV-positive adults. The impact of HIV-1 on Salmonella pathogenesis and the molecular basis for the differences between these bacteria and classical diarrhoeal S. Typhimurium remains unclear. Here, we
Laura Tedesco et al.
Diabetes, 59(11), 2826-2836 (2010-08-27)
Cannabinoid type 1 (CB1) receptor is involved in whole-body and cellular energy metabolism. We asked whether CB1 receptor stimulation was able to decrease mitochondrial biogenesis in different metabolically active tissues of obese high-fat diet (HFD)-fed mice. The effects of selective

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. I couldn't exactly understand what is that. Has it the same formulation with PBS?

    1 answer
    1. This product is made using potassium phosphate monobasic, but the pH is adjusted with NaOH, to pH ~7.4. This product is not phosphate buffered saline and the formulation is not the same.

      Helpful?

  4. gal = ガロン でしょうか

    1 answer
    1. Yes, gal represents gallon as the unit of measure. For this item, the '-1GA' is equivalent to 1 Gallon.

      Helpful?

  5. Could you share the composition of this product? Is it potassium or sodium?

    1 answer
    1. This product is prepared using potassium phosphate monobasic, but the pH is adjusted with NaOH, making this the mixed potassium/sodium salt.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service