Skip to Content
Merck
All Photos(1)

Key Documents

1805

Sigma-Aldrich

Phosphate Buffer

powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.47
Pricing and availability is not currently available.

form

powder

IVD

for in vitro diagnostic use

pH

7.60-7.80 (25 °C)

application(s)

hematology
histology

storage temp.

room temp

Components

Contains sodium and potassium phosphates. Intended for use in the Sigma Butyrate Esterase Procedure, No. 181B.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Romy Gleyse Chagas Barros et al.
Food research international (Ottawa, Ont.), 130, 108899-108899 (2020-03-12)
Edible flowers are receiving renewed interest as potential sources of bioactive compounds. The present study aimed to investigate the presence of bioactive compounds and antioxidant activity of some exotic flowers present in Brazil such as Amaranthus hypochondriacus, Tropaeolum majus (red)
Diya Wang et al.
Autophagy, 13(5), 914-927 (2017-03-21)
Central nervous system (CNS) inflammation and autophagy dysfunction are known to be involved in the pathology of neurodegenerative diseases. Manganese (Mn), a neurotoxic metal, has the potential to induce microglia-mediated neuroinflammation as well as autophagy dysfunction. NLRP3 (NLR family, pyrin
Ana-Marija Jagatić Korenika et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-26)
Commercial sparkling wine production represents a relatively low but important part of the Croatian wine production, especially in the Zagreb county. This study presents the results of volatile aroma compounds profile and organic acid composition of commercial sparkling wine samples

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service