Skip to Content
Merck
All Photos(1)

Key Documents

P3169

Sigma-Aldrich

pGlu-Phe-Leu p-nitroanilide

protease substrate, ≥98% (TLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H31N5O6
CAS Number:
Molecular Weight:
509.55
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

pGlu-Phe-Leu p-nitroanilide, protease substrate

Assay

≥98% (TLC)

form

powder

solubility

ethanol: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(Cc1ccccc1)NC(=O)C2CCC(=O)N2)C(=O)Nc3ccc(cc3)N(=O)=O

InChI

1S/C26H31N5O6/c1-16(2)14-21(25(34)27-18-8-10-19(11-9-18)31(36)37)29-26(35)22(15-17-6-4-3-5-7-17)30-24(33)20-12-13-23(32)28-20/h3-11,16,20-22H,12-15H2,1-2H3,(H,27,34)(H,28,32)(H,29,35)(H,30,33)

InChI key

NLVMZXVGZDVXDA-UHFFFAOYSA-N

Substrates

Chromogenic substrate for thiol proteases such as papain, ficin and bromelain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filippova IYu et al.
Analytical biochemistry, 143(2), 293-297 (1984-12-01)
L-Pyroglutamyl-L-phenylalanyl-L-leucine-p-nitroanilide (PFLNA)--a convenient chromogenic substrate for assay of thiol proteinases papain, ficin, and bromelain--was prepared by enzymatic synthesis with chymotrypsin as a catalyst. The thiol proteinases hydrolyze PFLNA with the liberation of p-nitroaniline, estimated spectrophotometrically by its absorbance at 410
Rim Hamza et al.
BMC plant biology, 18(1), 24-24 (2018-01-27)
Plants and insects have coexisted for million years and evolved a set of interactions which affect both organisms at different levels. Plants have developed various morphological and biochemical adaptations to cope with herbivores attacks. However, Tuta absoluta (Meyrick) (Lepidoptera: Gelechiidae)
Dalgys Martínez et al.
International journal of molecular sciences, 20(12) (2019-06-27)
The house dust mite (HDM) Dermatophagoides pteronyssinus is an important risk factor for asthma and rhinitis. Allergen specific immunotherapy that is based on recombinant proteins has been proposed for the safer and more efficient treatment of allergic diseases. The aim
Rim Hamza et al.
Bio-protocol, 8(17), e2993-e2993 (2018-09-05)
Enzymes play a key role in insect-plant relationships. For a better understanding of these interactions, we analyzed Tuta absoluta digestive enzymes. Here, we describe a detailed protocol for the detection of trypsin and papain-like enzymes in Tuta absoluta larvae by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service