Skip to Content
Merck
All Photos(1)

Key Documents

N8160

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide phosphate sodium salt

pkg of 10 mg (per vial)

Synonym(s):

β-NADP-Na, β-NADP-sodium salt, Coenzyme II sodium salt, NADP, TPN, TPN-Na, Triphosphopyridine nucleotide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H27N7O17P3 · Na
CAS Number:
Molecular Weight:
765.39
Beilstein:
4779954
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

form

powder

Quality Level

packaging

pkg of 10 mg (per vial)

mp

175-178 °C (dec.) (lit.)

solubility

water: 0.05 g/L

storage temp.

−20°C

SMILES string

[Na+].[H]O[H].NC(=O)c1ccc[n+](c1)C2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@@H]([C@@H]3O)P(O)(O)=O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H28N7O16P3.Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(45(33,34)35)14(30)11(43-21)6-41-47(38,39)44-46(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);;1H2/q;+1;/p-1/t10-,11-,13-,14-,15-,16-,20?,21-;;/m1../s1

InChI key

NLIGHACCLYZCOM-FHQWUTKFSA-M

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+) and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway (PPP).

Biochem/physiol Actions

Electron acceptor

Other Notes

Packaged based on NADP content as determined by UV-Vis.
This is the common form of NADP. Do not confuse with 3′-NADP or α-NADP.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bevan P Gang et al.
Cancer biology & therapy, 15(11), 1533-1541 (2014-12-09)
Deregulated metabolism is gaining recognition as a hallmark of cancer cells, and is being explored for therapeutic potential. The Warburg effect is a metabolic phenotype that occurs in 90% of tumors, where glycolysis is favored despite the presence of oxygen.
Rong Zhang et al.
American journal of respiratory and critical care medicine, 192(3), 315-323 (2015-05-07)
Lung-protective ventilatory strategies have been widely used in patients with acute respiratory distress syndrome (ARDS), but the ARDS mortality rate remains unacceptably high and there is no proven pharmacologic therapy. Mechanical ventilation can induce oxidative stress and lung fibrosis, which
Haiqin Chen et al.
Scientific reports, 5, 11247-11247 (2015-06-11)
Microorganisms are valuable resources for lipid production. What makes one microbe but not the other able to efficiently synthesize and accumulate lipids is poorly understood. In the present study, global gene expression prior to and after the onset of lipogenesis
María Apellániz-Ruiz et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(2), 322-328 (2014-11-16)
Paclitaxel, a widely used chemotherapeutic drug, can cause peripheral neuropathies leading to dose reductions and treatment suspensions and decreasing the quality of life of patients. It has been suggested that genetic variants altering paclitaxel pharmacokinetics increase neuropathy risk, but the
Dandan Liang et al.
Journal of molecular and cellular cardiology, 82, 63-74 (2015-03-12)
Diabetes mellitus is strongly associated with cardiomyopathy. The underlying mechanisms for the development of diabetic cardiomyopathy are complex and not completely understood. Recent studies showed that epidermal growth factor receptors (EGFRs) are involved in diabetes-induced cardiac injury. However, the role

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service