Skip to Content
Merck
All Photos(1)

Key Documents

N8035

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide phosphate sodium salt

pkg of 5 mg (per vial)

Synonym(s):

β-NADP-Na, β-NADP-sodium salt, Coenzyme II sodium salt, NADP, TPN, TPN-Na, Triphosphopyridine nucleotide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H27N7O17P3 · Na
CAS Number:
Molecular Weight:
765.39
Beilstein:
4779954
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

form

powder

packaging

pkg of 5 mg (per vial)

mp

175-178 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

[Na+].[H]O[H].NC(=O)c1ccc[n+](c1)C2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@@H]([C@@H]3O)P(O)(O)=O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H28N7O16P3.Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(45(33,34)35)14(30)11(43-21)6-41-47(38,39)44-46(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);;1H2/q;+1;/p-1/t10-,11-,13-,14-,15-,16-,20?,21-;;/m1../s1

InChI key

NLIGHACCLYZCOM-FHQWUTKFSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+) and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway (PPP).

Biochem/physiol Actions

Electron acceptor

Other Notes

Packaged based on NADP content as determined by UV-Vis.
This is the common form of NADP. Do not confuse with 3′-NADP or α-NADP.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuan Chen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 120, 116-125 (2017-09-10)
Water-soluble polymers are often used as pore formers to tailor permeability of film-forming hydrophobic polymers on coated dosage forms. However, their addition to a coating formulation could significantly increase the viscosity thus making the coating process difficult. Moreover, the dissolution
Bevan P Gang et al.
Cancer biology & therapy, 15(11), 1533-1541 (2014-12-09)
Deregulated metabolism is gaining recognition as a hallmark of cancer cells, and is being explored for therapeutic potential. The Warburg effect is a metabolic phenotype that occurs in 90% of tumors, where glycolysis is favored despite the presence of oxygen.
Rong Zhang et al.
American journal of respiratory and critical care medicine, 192(3), 315-323 (2015-05-07)
Lung-protective ventilatory strategies have been widely used in patients with acute respiratory distress syndrome (ARDS), but the ARDS mortality rate remains unacceptably high and there is no proven pharmacologic therapy. Mechanical ventilation can induce oxidative stress and lung fibrosis, which
Michael M Dubreuil et al.
Cell reports, 30(5), 1417-1433 (2020-02-06)
Reactive oxygen species (ROS) play critical roles in metabolism and disease, yet a comprehensive analysis of the cellular response to oxidative stress is lacking. To systematically identify regulators of oxidative stress, we conducted genome-wide Cas9/CRISPR and shRNA screens. This revealed a detailed
Paul A Fraser
Free radical biology & medicine, 51(5), 967-977 (2011-06-30)
The brain endothelium constitutes a barrier to the passive movement of substances from the blood into the cerebral microenvironment, and disruption of this barrier after a stroke or trauma has potentially fatal consequences. Reactive oxygen species (ROS), which are formed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service