Skip to Content
Merck
All Photos(5)

Key Documents

N0505

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate

Synonym(s):

Triphosphopyridine nucleotide sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7NaO17P3 · xH2O
CAS Number:
Molecular Weight:
765.39 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

chicken liver

Quality Level

Assay

≥98% (HPLC)
≥98% (spectrophotometric assay)

form

powder

mp

175-178 °C (dec.) (lit.)

solubility

H2O: soluble 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O)[C@H]([N+]2=CC(C(N)=O)=CC=C2)O[C@@H]1COP(OP(OC[C@H]3O[C@@H](N(C=N4)C5=C4C(N)=NC=N5)[C@H](OP(O)(O)=O)[C@@H]3O)([O-])=O)([O-])=O.O.[Na+]

InChI

1S/C21H28N7O17P3.Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);;1H2/q;+1;/p-1/t10-,11-,13-,14-,15-,16-,20-,21-;;/m1../s1

InChI key

CYQLUFJYVTUUFN-WUEGHLCSSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Electron acceptor

Packaging

Packaged by solid weight.

Other Notes

This is the common form of NADP. Do not confuse with 3′−NADP or α-NADP.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Moreno et al.
European journal of biochemistry, 161(3), 565-569 (1986-12-15)
Hexokinase PII is not inhibited by high Mg-ATP concentrations if the Mg2+-free ATP is kept at low levels (0.01 mM) in the assay mixture. Hexokinase PI activity is not affected either by Mg2+-free ATP nor by free Mg2+ in the
Syaghalirwa N M Mandiki et al.
Steroids, 70(2), 85-94 (2005-01-06)
Sex steroid inhibitors were used to characterize the effects of 17beta-estradiol (E2) and testosterone (T) on the sexual growth dimorphism of Eurasian perch juveniles. In experiment 1, growth responses to different doses of either E2 (25, 50, 75, and 100
David J Waxman et al.
Methods in molecular biology (Clifton, N.J.), 320, 91-96 (2006-05-25)
A spectrofluorometric method is described for the determination of CYP2A6-catalyzed coumarin 7-hydroxylation. Following acidification of the reaction mixture, the enzymatic product, 7-hydroxycoumarin, is recovered by a double-extraction procedure and assayed using an excitation wavelength of 370 nm and an emission
Considine, DM et al.
Van Nostrand Reinhold Encyclopedia of Chemistry, 1993-1993 null
J T Eppig et al.
Genetics, 103(4), 797-812 (1983-04-01)
Murine ovarian teratomas were used to determine recombination percentages for gene-gene and centromere-gene intervals. Data were obtained utilizing a recombinant inbred strain, LTXBJ, and a number of newly developed LT/SvEi congenic strains.--Centromere-gene recombination was measured at 11.3 +/- 1.2% for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service