MTOX1071
Kidney PTEC MDR1 KnockoutCells [MDR1 knock out (-/-)]
Human female kidney, proximal convoluted tubule epithelial cells
Sign Into View Organizational & Contract Pricing
All Photos(10)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.81
Recommended Products
Product Name
Kidney PTEC MDR1 KnockoutCells [MDR1 knock out (-/-)],
biological source
human female renal (proximal convoluted tubule epithelial cells)
form
liquid
OMIM accession no.
storage temp.
−196°C
Gene Information
human ... ABCB1(5243)
General description
Sigma modified human primary renal proximal tubule epithelial cells (RPTEC) with ZFNs to effectively create novel human RPTEC cell lines that can be utilized for investigating kidney toxicology (nephrotoxicity), transporters and various other renal studies. Sigma′s RPTEC′s exhibit normal epithelial morphology and expression of Renal Proximal Tubule (PT) specific markers such as, CD13 and α-GST. Sigma′s RPTEC′s maintain typical PT functions including albumin reabsorption, γ-glutamyl transpeptidase (GGT) activity, and respond to parathyroid hormone. The cells respond to several known human nephrotoxicants similar to human primary cells and demonstrate robust expression and activity of key kidney uptake and efflux transporters incuding OCT2, OATP4C1, OAT1, OCTN1, MRP2, MRP4, P-gp, MATE1, MATE2-K, PEPT1, and PEPT2.
Application
Human RPTEC SA7K control cell line and transporter KO models have been validated in several standard and 3D assays for measuring nephrotoxicity and transporter assays. RPTEC SA7K control cells can be used to measure toxicity of compounds using dose-response curves and viability such as MTT based in vitro Toxicology in both 2D and 3D culture.
RPTEC SA7K control cells can also be used to measure compound uptake/efflux by transporters using inhibitors. Precision editing in RPTEC SA7K transporter KO cells provides a more predictive platform for compound uptake/efflux by transporters without the need for non-specific inhibitors.
RPTEC SA7K cells have been validated for use on the Mimetas 3-lane OrganoPlate® in both toxicity and barrier integrity assays (( Z742750).
RPTEC SA7K cells can also be used on the AIM Biotech 3D cell culture chip in toxicity and permeability assays ( DAX01).
RPTEC SA7K control cells can also be used to measure compound uptake/efflux by transporters using inhibitors. Precision editing in RPTEC SA7K transporter KO cells provides a more predictive platform for compound uptake/efflux by transporters without the need for non-specific inhibitors.
RPTEC SA7K cells have been validated for use on the Mimetas 3-lane OrganoPlate® in both toxicity and barrier integrity assays (( Z742750).
RPTEC SA7K cells can also be used on the AIM Biotech 3D cell culture chip in toxicity and permeability assays ( DAX01).
Features and Benefits
Sigma′s human RPTEC control SA7K clone cells and knockout (KO) cells exhibit normal renal proximal tubule characteristics, functional renal uptake and efflux transporters and are sensitive to human nephrotoxicants making them ideal for studying drug-transporter interactions and early detection of renal toxicants. The SA7K clone should be used as a control for Sigma′s human RPTEC KO cells
- Sigma′s human RPTEC MDR1 (P-gp) Knockout (KO) allows investigations of drug transporter interactions involving P-gp in the kidney.
Quality
Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation
Other Notes
ADME/Tox cell lines license. These products are covered by the “Consumable Vial” License as described in Exhibit 1.
Legal Information
These products are covered by the ADME/Tox Cell Lines License as described in Exhibit 1, in the technical bulletin.
OrganoPlate is a registered trademark of MIMETAS B.V.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service