Skip to Content
Merck
All Photos(2)

Key Documents

MTOX1010

Sigma-Aldrich

HepaRG 5F Control Cells

human female liver (Source Disease: Hepatocarcinoma and Hepatitis C)

Synonym(s):

Human hepatocyte cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.24

product name

HepaRG 5F Control Cells, 1 vial

biological source

human female liver (Source Disease: Hepatocarcinoma and Hepatitis C)

form

liquid

storage temp.

−196°C

General description

This product consists of ZFN engineered HepaRG 5F clone control cells. They are intended for use as both control cells for HepaRG knockout cells as well as for a wide variety of liver cell based assays. Hepa RG cells display hepatocyte-like functions and can be a replacement to primary human hepatocytes in a number of drug metabolism, disposition and genotoxicity studies.
HepaRG is a human hepatoma cell line. The cells possess a pseudodiploid karyotype and have been characterized as an oval ductular bipotent hepatic cell line as they have the ability to differentiate into both biliary and hepatocyte lineages in the presence of DMSO. HepaRG cells express the major xenobiotic sensors (PXR, CAR and AhR), drug transporters, phase I and II drug metabolizing enzymes as well as key hepatic transcription factors involved in stress response pathways.

Application

HepaRG 5F Control Cells has been used in a gene expression assay to study the effect of pregnane X receptor on the induction of human cytochrome P450 3A4 by the irreversible myeloperoxidase inactivator PF-06282999.
See technical bulletins for detailed protocols

Features and Benefits

Sigma′s HepaRG 5F Clone exhibits improved growth characteristics and enhanced functionality, expressing a large panel of drug metabolism enzymes; including cytochrome P450 enzymes CYP1A2, CYP2B6, CYP2C8/9, CYP2C19, CYP2D6, CYP2E1, and CYP3A4, nuclear receptors AhR, PXR, CAR, LXR, and FXR, and phase II enzymes UGT, SULT, NAT and GST.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation

Legal Information

These products are covered by the License Agreement as described in Exhibit 1 and 2, in the technical bulletin.
HepaRG is a trademark of BioPredic International company

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The HepaRG cell line: a unique in vitro tool for understanding drug metabolism and toxicology in human.
Andersson, Tommy
Expert Opinion on Drug Metabolism & Toxicology, 8, 909-920 (2012)
Induction of human cytochrome P450 3A4 by the irreversible myeloperoxidase inactivator PF-06282999 is mediated by the pregnane X receptor.
Moscovitz, Jamie E., et al.
Xenobiotica, 1-9 (2017)
Assessment of the genotoxic potential of indirect chemical mutagens in HepaRG cells by the comet and the cytokinesis-block micronucleus assays.
Le Hegarat, Ludovic, et al.
Mutagenesis, 25, 555-560 (2010)
Tristan M Sissung et al.
Molecular pharmacology, 96(2), 158-167 (2019-06-09)
Mithramycin demonstrates preclinical anticancer activity, but its therapeutic dose is limited by the development of hepatotoxicity that remains poorly characterized. A pharmacogenomics characterization of mithramycin-induced transaminitis revealed that hepatotoxicity is associated with germline variants in genes involved in bile disposition:
Khristy J Thompson et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 32(6), 829-843 (2019-09-22)
Manganese (Mn) is an essential element necessary for proper development and brain function. Circulating Mn levels are regulated by hepatobiliary clearance to limit toxic levels and prevent tissue deposition. To characterize mechanisms involved in hepatocyte Mn uptake, polarized human HepaRG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service