Skip to Content
Merck
All Photos(1)

Documents

G7513

Sigma-Aldrich

L-Glutamine solution

200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture

Synonym(s):

Glavamin, Levoglutamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

300
400

product line

BioXtra

form

solution

concentration

200 mM

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

shipped in

dry ice

storage temp.

−20°C

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

L-glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Application

L-glutamine is an essential amino acid that is a crucial component of culture media, which serves as a major energy source for cells in culture. Optimal cell performance usually requires supplementation of the medium with L-glutamine prior to use.

Preparation Note

Prepared in cell culture grade water.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ofir Cohn et al.
Scientific reports, 6, 37115-37115 (2016-11-16)
SETD3 is a member of the protein lysine methyltransferase (PKMT) family, which catalyzes the addition of methyl group to lysine residues. Accumulating data suggest that PKMTs are involved in the regulation of a broad spectrum of biological processes by targeting
Miguel Pérez-Garrastachu et al.
Nucleus (Austin, Tex.), 8(5), 515-533 (2017-07-12)
Nucleoporins are the main components of the nuclear-pore complex (NPC) and were initially considered as mere structural elements embedded in the nuclear envelope, being responsible for nucleocytoplasmic transport. Nevertheless, several recent scientific reports have revealed that some nucleoporins participate in
Boris Redko et al.
Oncotarget, 8(1), 757-768 (2016-10-22)
The newly discovered short (9 amino acid) non-RGD S-S bridged cyclic peptide ALOS-4 (H-cycl(Cys-Ser-Ser-Ala-Gly-Ser-Leu-Phe-Cys)-OH), which binds to integrin αvβ3 is investigated as peptide carrier for targeted drug delivery against human metastatic melanoma. ALOS4 binds specifically the αvβ3 overexpressing human metastatic
Zlata Vershinin et al.
The Journal of biological chemistry, 291(13), 6786-6795 (2016-02-05)
Lysine methylation of non-histone proteins has emerged as a key regulator of many cellular functions. Although less studied than other post-translational modifications such as phosphorylation and acetylation, the number of known methylated non-histone proteins is rapidly expanding. We have identified
Nara Shin et al.
Environmental science. Processes & impacts, 21(8), 1342-1352 (2019-05-03)
In this study, we investigated the airborne particles released during paper printing and paper shredding processes in an attempt to characterize and differentiate these particles. Particle characteristics were studied with real time instruments (RTIs) to measure concentrations and with samplers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service