Skip to Content
Merck
All Photos(1)

Key Documents

MSKT0002

Sigma-Aldrich

SOLu-Trypsin Rapid Digestion Kit

Proteomics grade

Synonym(s):

Rapid Trypsin, Recombinant Trypsin for Protein Digestion in Mass Spectrometry, rTrypsin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331
NACRES:
NA.78

grade

Proteomics Grade

form

solution

suitability

suitable for mass spectrometry

shipped in

wet ice

storage temp.

2-8°C

General description

SOLu-Trypsin Rapid Digestion Kit (MSKT0002) consists of 400 ug of solution-stable trypsin (EMS0004) suitable for protein sequencing, and 30 mL of Rapid Digestion Buffer (EMS0009), allowing for trypsin digestion in one hour at 60°C. Trypsin should be used at a 1:10 enzyme:substrate ratio in Rapid Digestion Buffer, which can be substituted into existing user workflows.

Features and Benefits

  • Provides a fast and efficient method for protein digestion.
  • Removal of denaturants by offline desalting is not necessary, which further shortens the time for sample preparation.
  • Compatible with a variety of sample types, reduction and alkylation protocols, and downstream workflows.

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service