Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0851-01

Disposable PD 10 Desalting Columns

Cytiva 17-0851-01, pack of 30 pieces

Synonym(s):

Disposable desalting columns, PD-10 columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

feature

wetted part: no

packaging

pack of 30 pieces

manufacturer/tradename

Cytiva 17-0851-01

storage condition

dry media

bed size

14.5 mm × 50 mm

bed volume

8.3 mL

column I.D.

15 mm

matrix

cross-linked dextran

particle size

>50 μm (dry)
85-260 μm

cleaning

2-13

working range

2-13

suitability

suitable for bioprocess medium

Related Categories

General description

Disposable PD-10 Desalting Columns offer an inexpensive and convenient alternative to lengthy and troublesome dialysis procedures. In minutes, biological samples are desalted and small molecular weight solutes are removed or exchanged with excellent recoveries of macromolecules.

The instructions for Disposable PD-10 Desalting Columns have been updated with two new application protocols: gravity or centrifugation, to increase the flexibility of the product. The gravity protocol allows a simple cleanup of samples with one or several columns in parallel, without the need for a purification system. The spin protocol enables runs in parallel in a standard centrifuge with minimal dilution of the eluted sample.

Four adapters are included in each product pack to enable easy centrifugation. Together with LabMate PD-10 Buffer Reservoir, wash and equilibration buffer can be applied in one step with the gravity flow protocol. Empty PD-10 columns are recommended for use with bulk media for affordable and versatile sample preparation.

Features and Benefits

  • Fast, affordable, high-recovery desalting and buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex® G-25 Medium.
  • Flexible protocols allow runs by gravity flow or centrifugation.
  • Easy to use and equilibrate; apply sample; elute.
  • Preparation of the samples in the range of 1.0 to 2.5 mL by gravity flow and 1.75 to 2.5 mL with centrifugation.
  • Desalt multiple samples in parallel with a desalting capacity >90%.
  • Recovery in the range 70 to >95%.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.15% Kathon CG/ICP Biocide)
Store at 4 to 30 °C (4 to 8, Used Media 20% Ehtanol or 0.01 M NaOH)

Legal Information

Sephadex is a registered trademark of Cytiva

also commonly purchased with this product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye Guan et al.
eLife, 7 (2018-04-06)
Single-cell analysis is pivotal to deciphering complex phenomena like heterogeneity, bistability, and asynchronous oscillations, where a population ensemble cannot represent individual behaviors. Bulk cell-free systems, despite having unique advantages of manipulation and characterization of biochemical networks, lack the essential single-cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service