Skip to Content
Merck
All Photos(1)

Key Documents

M9917

Sigma-Aldrich

Mannan−Agarose

saline suspension

Synonym(s):

Mannan Binding Protein Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

biological source

plant

form

saline suspension

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

hydroxyl

matrix spacer

1 atom

capacity

≥30 mg/mL binding capacity (concanavalin A)

suitability

suitable for chromatography

storage temp.

2-8°C

Application

Mannan-agarose is used in protein chromatography, affinity chromatography and carbohydrate matrices. Mannan-agarose has been used in studies assessing poly(ADP-ribosyl)ation of mannose-binding lectin out of human kidney cells and to inform mannose-associated serine protease (MASP) activated components of the coagulation system. Mannan-agarose has also been used to study the innate immunity and resistance to Aeromonas salmonicida exhibited by the sea lamprey (Petromyzon marinus), a parasitic cartilaginous fish of the North American Great Lakes and a predator of many bony fish species of commercial importance to the fishing industry. Mannan-Agarose has been used to deplete rabbit anti-mannose-binding lectin (MBL) polyclonal antiserum of anti-mannan antibodies.

Physical form

Suspension in 1.0 M NaCl, 10 mM imidazole containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donald D Ourth et al.
Fish & shellfish immunology, 23(6), 1152-1160 (2007-10-24)
Mannose-binding C-type lectin (MBL) was isolated from channel catfish (Ictalurus punctatus) NWAC 102 and 103 strains, blue catfish (Ictalurus furcatus) D+B and Rio Grande strains, hybrid catfish (channel catfish female NWAC 103 x blue catfish male D+B) sera, and purified
M C Montalto et al.
Journal of immunology (Baltimore, Md. : 1950), 166(6), 4148-4153 (2001-03-10)
Complement plays a significant role in mediating endothelial injury following oxidative stress. We have previously demonstrated that the lectin complement pathway (LCP), which is initiated by deposition of the mannose-binding lectin (MBL), is largely responsible for activating complement on endothelial
Kazuhiko Takahara et al.
International immunology, 16(6), 819-829 (2004-04-21)
The mouse (m) DC-SIGN family consists of several homologous type II transmembrane proteins located in close proximity on chromosome 8 and having a single carboxyl terminal carbohydrate recognition domain. We first used transfected non-macrophage cell lines to compare the polysaccharide
Human follicular lymphoma cells contain oligomannose glycans in the antigen-binding site of the B-cell receptor
Radcliffe CM, et al.
The Journal of Biological Chemistry (2007)
T Kawai et al.
Glycobiology, 8(3), 237-244 (1998-05-16)
Mannan-binding protein (MBP) is a member of the collectin family of protein. There are two types of MBP, MBP-A and MBP-C, which were found in rodent (rats and mice), rhesus monkey, and cynomolgus monkey, while chimpanzee and human have only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service