Skip to Content
Merck
All Photos(1)

Key Documents

M6400

Sigma-Aldrich

D-Mannose Agarose

saline suspension

Synonym(s):

D-Mannose resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

biological source

birch/beech wood hydrolysate

form

saline suspension

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

matrix activation

divinyl sulfone

matrix attachment

hydroxyl

matrix spacer

5 atoms

capacity

≥40 mg/mL binding capacity (concanavalin A)

suitability

suitable for chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Mannose-agarose is used in protein chromatography, affinity chromatography and carbohydrate matrices. Mannose-agarose has been used in studies assessing poly(ADP-ribosyl)ation of mannose-binding lectin out of human kidney cells and to inform mannose-associated serine protease (MASP) activated components of the coagulation system. Mannose-agarose has also been used to study the innate immunity and resistance to Aeromonas salmonicida exhibited by the sea lamprey (Petromyzon marinus), a parasitic cartilaginous fish of the North American Great Lakes and a predator of many bony fish species of commercial importance to the fishing industry.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Yagi et al.
Plant & cell physiology, 37(7), 1007-1012 (1996-10-01)
A mannose-specific lectin was isolated from leaves of Neoregelia flandria, an ornamental plant that belongs to Bromeliaceae, a family of monocotyledons. The amino acid composition and molecular mass of the lectin were similar to those of mannose-specific lectins from other
E N Kaufman et al.
Biophysical journal, 58(4), 873-885 (1990-10-01)
Fluorescence Recovery After Photobleaching (FRAP) has been used extensively in the study of transport and binding in biological media in vitro. The present study adapts and further develops FRAP so that it may be utilized for the in vivo quantification
Donald D Ourth et al.
Veterinary immunology and immunopathology, 126(3-4), 407-412 (2008-10-22)
The sea lamprey (Petromyzon marinus) is a parasitic cartilaginous fish of the North American Great Lakes and a predator of many bony fish species of commercial importance to the fishing industry. Mannose-binding C-type lectin (MBL) was isolated by mannan-agarose affinity
Momoe Takahashi et al.
Journal of immunology (Baltimore, Md. : 1950), 176(8), 4861-4868 (2006-04-06)
The recognition of pathogens is mediated by a set of pattern recognition molecules that recognize conserved pathogen-associated molecular patterns shared by broad classes of microorganisms. Mannose-binding lectin (MBL) is one of the pattern recognition molecules and activates complement in association
H Ushijima et al.
Japanese journal of cancer research : Gann, 83(5), 458-464 (1992-05-01)
The lectin-like protein analogous to bovine conglutinin was purified from human serum. The carbohydrate-binding ability of conglutinin-like protein was inhibited by D-mannose, N-acetylglucosamine and L-fucose as well as by mannan-containing oligosaccharides. By applying a lectin-based ELISA system it was demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service