M9062
Greiner CELLSTAR® multiwell culture plates
6 wells (suspension culture with lid)
Synonym(s):
culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
material:
flat clear wells
polystyrene
polystyrene
size:
6 wells
sterility:
sterile
binding type:
non-treated surface
feature:
lid
Recommended Products
material
flat clear wells
polystyrene
sterility
sterile
feature
lid
packaging
pack of 1 ea
manufacturer/tradename
Greiner 657185
size
6 wells
well volume
16 mL
well working volume
2- 5 mL
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The Greiner CELLSTAR® multiwell culture plate is a 6-well polystyrene plate. It is designed for cell culture. Cell Culture Multiwell Plates has a lid with condensation rings. Each plate is sterile and single-packed for easy use.
Application
Greiner CELLSTAR® multiwell culture plates have been used:
- to prepare PANC1 human pancreatic duct cell line/human islet cells and islet-derived progenitor cells (hIPCs) for puromycin selection of transduced cells.
- for fixed z-direction (FiZD) kidney primordia culture to be used in time-lapse confocal imaging.
- for the cultivation of corneal epithelialcells.
Features and Benefits
- With hydrophobic surface for suspension culture cells and hybridoma cells
- Free of detectable DNase, RNase, human DNA
- Preserved cell viability and integrity
- Single position lids to prevent cross-contamination
- High clarity and low autofluorescence for clear observation of cell cultures
- Contains single position lids to prevent cross-contamination
- Lid enables optimal gas exchange with minimal evaporation
- Alphanumeric well coding for easy identification and tracking
- Seamless integration with automated systems
- Convenient, easy-to-open packaging
- Working volume per well: 16 ml
- Sterile and non-pyrogenic
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Alice Rocha Teixeira Netto et al.
Methods and protocols, 6(3) (2023-05-23)
In ophthalmic research, there is a strong need for in vitro corneal cell models. Here, we describe different protocols for the cultivation of primary corneal cells that were isolated from porcine eyes. This primary cell culture can be used to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service