Skip to Content
Merck
All Photos(1)

Key Documents

M8687

Sigma-Aldrich

Greiner CELLSTAR® multiwell culture plates

Greiner CELLSTAR® multiwell culture plates
1 of 1 reviewers received a sample product or took part in a promotion

12 wells (TC treated with lid), flat bottom, pack of 100 ea sterile, individually wrapped

Synonym(s):

culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
flat bottom
flat bottom wells
polystyrene
base surface area × working volume:
3.9 cm2 × 2-4 mL
size:
12 wells
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
Pricing and availability is not currently available.

material

flat bottom
flat bottom wells
polystyrene

sterility

sterile

feature

lid

packaging

pack of 100 ea sterile, individually wrapped

manufacturer/tradename

Greiner 665180

base surface area × working volume

3.9 cm2 × 2-4 mL

size

12 wells

color

clear

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Greiner CELLSTAR® multiwell culture plates have been used to culture the cells for quantitative polymerase chain reaction (qPCR) and western blotting. It has also been used to perform infectious viral load assessment by plaque assay.

Legal Information

CELLSTAR is a registered trademark of Greiner Bio-One GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service