Skip to Content
Merck
All Photos(5)

Key Documents

M9024

Sigma-Aldrich

MOPS sodium salt

BioPerformance Certified, suitable for cell culture, ≥99.5%

Synonym(s):

3-(N-Morpholino)propanesulfonic acid sodium salt, 4-Morpholinepropanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14NNaO4S
CAS Number:
Molecular Weight:
231.25
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

BioPerformance Certified

Assay

≥99.5%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and bioburden, tested

useful pH range

6.5-7.9

pKa (25 °C)

7.2

application(s)

diagnostic assay manufacturing

foreign activity

DNase, RNase, NICKase and protease, none detected

SMILES string

[Na+].[O-]S(=O)(=O)CCCN1CCOCC1

InChI

1S/C7H15NO4S.Na/c9-13(10,11)7-1-2-8-3-5-12-6-4-8;/h1-7H2,(H,9,10,11);/q;+1/p-1

InChI key

MWEMXEWFLIDTSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Buffers more than buffering agent: introducing a new class of stabilizers for the protein BSA.: This research highlights the use of MOPS sodium salt as a novel stabilizing agent for bovine serum albumin (BSA), extending its role beyond a simple buffering agent. The study demonstrates how MOPS sodium salt can improve the stability of BSA under various conditions, providing significant insights for biochemical applications and protein preservation (Gupta BS et al., 2015).


Other Notes

Easily compare specifications for MOPS sodium salt products with the MOPS sodium salt specification table.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Hou et al.
Biotechnology progress, 31(4), 974-982 (2015-05-29)
Protein A chromatography is widely employed for the capture and purification of monoclonal antibodies (mAbs). Because of the high cost of protein A resins, there is a significant economic driving force to seek new downstream processing strategies. Membrane chromatography has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service