Skip to Content
Merck
All Photos(5)

Key Documents

M9381

Sigma-Aldrich

MOPS sodium salt

≥99.5% (titration)

Synonym(s):

3-(N-Morpholino)propanesulfonic acid sodium salt, 4-Morpholinepropanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14NNaO4S
CAS Number:
Molecular Weight:
231.25
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥99.5% (titration)

form

powder

useful pH range

6.5-7.9

pKa (25 °C)

7.2

solubility

water: 33 % (w/w), clear, colorless

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

[Na+].[O-]S(=O)(=O)CCCN1CCOCC1

InChI

1S/C7H15NO4S.Na/c9-13(10,11)7-1-2-8-3-5-12-6-4-8;/h1-7H2,(H,9,10,11);/q;+1/p-1

InChI key

MWEMXEWFLIDTSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Buffers more than buffering agent: introducing a new class of stabilizers for the protein BSA.: This research highlights the use of MOPS sodium salt as a novel stabilizing agent for bovine serum albumin (BSA), extending its role beyond a simple buffering agent. The study demonstrates how MOPS sodium salt can improve the stability of BSA under various conditions, providing significant insights for biochemical applications and protein preservation (Gupta BS et al., 2015).


Packaging

The DRYP®OUR®packaging system for high-quality hygroscopic salts DRYPOUR®packaging:
  • The current DRYPOUR®packaging configuration is in a 50L nestable drum
  • 25KG-DP are the standard prepack offering
  • Smaller volumes can be put in the same drum for variable configurations
The benefits of DRYPOUR®:
  • Improves operational efficiency by reducing material preparation time and increasing operator safety
  • Minimizes caking of hygroscopic salts
  • Offers triple protection: against moisture inside and outside, and against contamination
How does the DRYPOUR®packaging system Improve your operational efficiency?
  • This state-of-the-art design packaging system minimizes caking, thus significantly reducing the time needed for material preparation
  • PE drum with our new tamper-evident seal, a polyethylene liner with integrated desiccant bags and a breathable interior Tyvek®liner
  • Desiccant bags are firmly integrated into the polyethylene liner
  • Triple protection delivers two invaluable results:
  1. Dramatically reduced caking
  2. No contamination risk from the desiccant

Other Notes

Easily compare specifications for MOPS sodium salt products with the MOPS sodium salt specification table.

Legal Information

DRYPOUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emmanuel Uchenna Ahiwe et al.
Asian-Australasian journal of animal sciences, 33(4), 579-587 (2019-09-05)
This study evaluated the effect of yeast products on growth performance, visceral organ weights, endogenous enzyme activities, ileal nutrient digestibility and meat yield of broiler chickens fed diets containing autolyzed whole yeast (WY) and yeast cell walls (YCW) at varying
Ying Hou et al.
Biotechnology progress, 31(4), 974-982 (2015-05-29)
Protein A chromatography is widely employed for the capture and purification of monoclonal antibodies (mAbs). Because of the high cost of protein A resins, there is a significant economic driving force to seek new downstream processing strategies. Membrane chromatography has

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Hi I was wondering if this compound MOPs free acid and the MOPS Sodium Salt have an expiry date?

    1 answer
    1. These products are not assigned an expiration date. They is assigned a recommended retest date of 2 years after the quality release date. A recommended retest date is the period of time during which the products are expected to remain within established stability specifications, provided that they has been stored under defined conditions. After the Retest Date, samples should be examined to ensure that the products are still in compliance with the established specifications. For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service