Skip to Content
Merck
All Photos(1)

Key Documents

475898

Millipore

MOPS

≥99% (titration), solid, Ultrol® Grade

Synonym(s):

MOPS, Free Acid, ULTROL® Grade, 3-(N-Morpholino)propanesulfonic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO4S
CAS Number:
Molecular Weight:
209.26
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.25

Product Name

MOPS, ULTROL® Grade, ≥99% (titration), zwitterionic buffer useful in pH range 6.5-7.9

description

Merck USA index - 14, 6265

Quality Level

Assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

7.2

solubility

water: 1 M

cation traces

heavy metals: ≤1 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])CCC[N+H]1CCOCC1

InChI

1S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)

InChI key

DVLFYONBTKHTER-UHFFFAOYSA-N

General description

A zwitterionic buffer useful in the pH range of 6.5-7.9. Has a pKa of 7.20 at 25°C.
Absorbance (1.0 M, H2O, 260 nm):≤0.05.
One of the zwitterionic buffers active in the pH range of 6.5 to 8.0 (absorbance: ≤0.05 at 260 nm, 1.0 M, H2O). Has a pK,sub>a of 6.95-7.24 at 20°C.

Warning

Toxicity: Standard Handling (A)

Other Notes

Spence, J.T., et al. 1988. Biochem. J. 250, 921.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service