Skip to Content
Merck
All Photos(1)

Documents

A2278

Sigma-Aldrich

N-Acetyl-D-glucosamine–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
PubChem Substance ID:
NACRES:
NA.56

form

saline suspension

matrix

6% beaded agarose

matrix activation

divinyl sulfone

matrix attachment

hydroxyl

matrix spacer

5 atoms

capacity

≥25 mg/mL binding capacity (Triticum vulgaris lectin)

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C8H15NO6/c1-4(12)9-5(2-10)7(14)8(15)6(13)3-11/h2,5-8,11,13-15H,3H2,1H3,(H,9,12)/t5-,6+,7+,8+/m0/s1

InChI key

MBLBDJOUHNCFQT-LXGUWJNJSA-N

General description

N-Acetyl-D-glucosamine is used in affinity chromatography, protein chromatography, and carbohydrate matrices. N-Acetyl-D-glucosamine can be used to purify wheat germ (Triticum vulgaris) lectin.

Application

N-Acetyl-D-glucosamine-Agarose has been used

  • for the incubation of supernatant for the depletion of ficolin-3-deficient serum
  • for detecting the lectin activity by ligand binding assays
  • to produce ficolin 2 depleted serum

Physical form

Suspension in 0.15 M NaCl, 0.01 M sodium phosphate, pH 6.8, containing 0.02% sodium azide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M B Allen et al.
The Biochemical journal, 185(3), 577-582 (1980-03-01)
1. Under normal assay conditions the N-acetyl-D-glucosamine kinases from rat liver and kidney show a pH-dependent lag phase before reaching a steady state, which is probably due to reversible dissociation of the dimeric enzyme. 2. The enzyme catalyses the phosphorylation
R Sullivan et al.
Biochemical and biophysical research communications, 162(1), 184-188 (1989-07-14)
Human ejaculated spermatozoa are heterogeneous and can be separated into two distinct populations according to their respective buoyant densities. In order to investigate the functional differences between these two types of spermatozoa, we have searched for the presence of galactosyltransferase.
J B Pitner et al.
Carbohydrate research, 324(1), 17-29 (2000-03-21)
The binding of Strep 9, a mouse monoclonal antibody (mAb) of the IgG3 subclass directed against the cell-wall polysaccharide of Group A Streptococcus (GAS), has been characterized. The intact antibody and proteolytic fragments of Strep 9 bind differently to GAS:
L Munthe-Fog et al.
Scandinavian journal of immunology, 65(4), 383-392 (2007-03-28)
Ficolin-2 (L-ficolin), derived from the FCN2 gene, is an innate immunity pattern recognition molecule found in human serum in which inter-individual variation in serum appears to be under genetic control. To validate and extend this finding, we developed a sandwich
M B Allen et al.
The Biochemical journal, 185(3), 565-575 (1980-03-01)
1. Procedures for the extensive purification in high yield of N-acetyl-D-glucosamine kinase from rat liver and kidney are described. The separation of this enzyme from hepatic glucokinase depended primarily on their differing behaviour on an affinity column of Sepharose--N-(6-aminohexanoyl)-2-amino-2-deoxy-D-glucopyranose. 2.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service