LH20100
Sephadex® LH-20 Size Exclusion Resin
140-550 mesh
Synonym(s):
Lipophilic Sephadex®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
13111000
NACRES:
NB.21
Recommended Products
Product Name
Sephadex® LH-20,
form
slurry
technique(s)
LPLC: suitable
matrix
Agarose
matrix active group
polymer
swelling
1 g swells to 4 mL (water, methanol)
bead size
25-100 μm
operating pH range
2-13
separation technique
size exclusion (SEC)
storage temp.
room temp
Related Categories
Application
Sephadex® LH-20 is a crosslinked dextran-based resin used with organic solvent mobile phases for gel permeation, normal phase partition, and adsorption chromatography of lipids, steroids, fatty acids, hormones, vitamins, etc. It is also used in gel filtration chromatography, protein chromatography, gel filtration media, resins and separation media.
Sephadex® LH-20 has been used to isolate bioactive polyphenolic content from blackberry genotypes and tryptophan from the aqueous extract of lotus rhizomes.
Sephadex® LH-20 has been used to isolate bioactive polyphenolic content from blackberry genotypes and tryptophan from the aqueous extract of lotus rhizomes.
Legal Information
Sephadex is a registered trademark of Cytiva
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yizhong Cai et al.
Journal of chromatographic science, 43(9), 454-460 (2005-10-11)
HPLC characterization of reversed-phase (RP) high-performance liquid chromatography (HPLC) has been widely used in separation and identification of plant pigments. An effective RP-HPLC-based method is established to systematically isolate, identify, and quantitate the betalain pigments in the plants of 37
S K Kundu et al.
Journal of lipid research, 20(7), 825-833 (1979-09-01)
Aminopropyl silica gel was prepared from porous silica gel and was used as a solid support for immunoadsorbent in the purification of anti-glycolipid antibodies. For neutral glycosphingolipids, a carboxyl function was generated by oxidation of the olefinic double bond of
Suman B Sharma et al.
Journal of medicinal food, 14(4), 353-359 (2011-03-05)
Diabetes is accompanied by lipid abnormalities, which contribute significantly to cardiovascular morbidity and mortality in diabetic patients. We previously demonstrated the potent antihyperglycemic activity of the active principle (fraction II from Sephadex LH 20 chromatography [LH II]) isolated from ethanolic
Encapsulated three-dimensional culture supports development of nonhuman primate secondary follicles.
Min Xu et al.
Biology of reproduction, 81(3), 587-594 (2009-05-29)
In vitro ovarian follicle cultures may provide fertility-preserving options to women facing premature infertility due to cancer therapies. An encapsulated three-dimensional (3-D) culture system utilizing biomaterials to maintain cell-cell communication and support follicle development to produce a mature oocyte has
A Cahouet et al.
International journal of pharmaceutics, 242(1-2), 367-371 (2002-08-15)
The aim of the present work was to study the biodistribution of a radiolabeled lipidic nanocapsule formulation after intravenous administration in rat by scintigraphy and gamma counting. This formulation is expected to be used as anticancer agent delivery devices and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service