Skip to Content
Merck
All Photos(2)

Key Documents

G2580

Sigma-Aldrich

Sephadex® G-25

Fine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56
Pricing and availability is not currently available.

Quality Level

technique(s)

buffer exchange: suitable

matrix active group

phase

swelling

1 g swells to 4-6 mL

bead size

20-80 μm

application(s)

life science and biopharma

compatibility

Cytiva

InChI

1S/C6H12O6.C3H8O3/c7-1-2-3(8)4(9)5(10)6(11)12-2;4-1-3(6)2-5/h2-11H,1H2;3-6H,1-2H2/t2-,3-,4-,5-,6-;/m1./s1

InChI key

BCPVZFFJGCURNR-OCOFDJSDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sephadex® is a beaded gel filtration medium prepared by cross-linking dextran with epichlorohydrin under alkaline conditions. It is used in affinity chromatography, protein chromatography and gel filtration chromatography.
Fractionation Range (MW)
Globular Proteins: 1,000 - 5,000
Dextrans: 100 - 5,000

Application

Sephadex® G-25 has been used in size-exclusion chromatography to remove excess dye from labelled proteins.[1]

Other Notes

G2580-100G′s updated product number is GE17-0032-01
G2580-500G′s updated product number is GE17-0032-02

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Celine Hoffmann et al.
Analytical biochemistry, 372(1), 62-71 (2007-09-11)
Fluorescein isothiocyanate (FITC) is a well-known probe for labeling biologically relevant proteins. However, the impact of the labeling procedure on protein structure and biological activities remains unclear. In this work, FITC-labeled human plasma fibronectin (Fn) was developed to gain insight
Jana Schwarzer et al.
Electrophoresis, 29(20), 4203-4214 (2008-10-18)
Glycoproteins, such as monoclonal antibodies as well as recombinant and viral proteins produced in mammalian cell culture play an important role in manufacturing of many biopharmaceuticals. To ensure consisting quality of the corresponding products, glycosylation profiles have to be tightly
Conformations of a metastable SH3 domain characterized by smFRET and an excluded-volume polymer model.
Mazouchi A, et al.
Biophysical Journal, 110(7), 1510-1522 (2016)
Jennifer Belyea et al.
Biochemistry, 45(48), 14275-14284 (2006-11-30)
The study of axial ligation by anionic ligands to ferric heme iron by resonance Raman spectroscopy provides a basis for comparison of the intrinsic electron donor ability of the proximal histidine in horse heart myoglobin (HHMb), dehaloperoxidase (DHP), and horseradish
P M G Paiva et al.
Bioresource technology, 88(1), 75-79 (2003-02-08)
A highly purified trypsin inhibitor was obtained from Echinodorus paniculatus when an extract prepared from E. paniculatus seed flour (25 gl(-1), with 0.1 M ammonium acetate buffer, pH 8.3, under agitation for 6 min at 28 degrees C) was chromatographed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service