Skip to Content
Merck
All Photos(1)

Key Documents

13606

Supelco

Diaion® HP-20

pkg of 500 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817

form

solid

Quality Level

packaging

pkg of 500 g

technique(s)

HPLC: suitable
LPLC: suitable
ion chromatography: suitable
thin layer chromatography (TLC): suitable

surface area

~500 m2/g

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

250-850 μm

pore size

~1.30 mL/g pore volume
260 Å mean pore size

density

1.01 g/mL at 25 °C (true wet)(lit.)

separation technique

reversed phase

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diaion® HP-20 is a non-polar copolymer styrene-divynilbenzene adsorbent resin. It is a polymeric adsorbent used in reverse phase chromatography. It has spherical particles with 0.5mm diameter. Diaion HP-20 has been considered as the most effective adsorbent to eliminate toxic effects in fermentation. It is a synthetic adsorbent used for preconcentration and separation of traces heavy metal ions.

Application

  • Diaion® HP-20 was used as adsorbent for open column chromatography.
  • Diaion® HP-20 was used in purification of glucosides using TLC and HPLC analysis.
Polyaromatic adsorbent resin for hydrophobic compounds: antibiotics, biomolecules; useful for desalting; broad application base.

Other Notes

Swelling in toluene = 30%

Legal Information

Diaion is a registered trademark of Mitsubishi Chemical Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of Reverse Phase SMB-Chromatographic Bioseparations of Amino Acid Aqueous Solutions.
Molnar, Z., et al
Hungarian Journal of Industrial Chemistry, 32.1, 23-23 (2004)
J C Lee et al.
Letters in applied microbiology, 37(3), 196-200 (2003-08-09)
To use adsorbent resins in fermentations to eliminate toxic effects on growth, reduce feedback repression of production and assist in recovery of teicoplanin. An adsorbent resin was added to the culture broth for the adsorption of teicoplanin. Amberlite XAD-16, Diaion
B Jiang et al.
Journal of ethnopharmacology, 96(3), 521-528 (2004-12-28)
Ethnopharmacological collections of plants used in traditional medical systems are a valuable but often underappreciated resource for scientific investigation. These collections contain many samples of plants currently employed in herbal and pharmaceutical medicine, and questions on stability and storage life
Hiroaki Inoue et al.
Applied and environmental microbiology, 68(10), 4809-4811 (2002-09-27)
The addition of HP-20 resin to a medium could enhance the growth of Legionella species. Elemental sulfur was isolated as a self-growth-inhibiting substance produced endogenously by Legionella pneumophila from methanol extracts of the resins used to culture the bacterium. Elemental
S Irmler et al.
The Plant journal : for cell and molecular biology, 24(6), 797-804 (2001-01-03)
The molecular characterization of CYP72A1 from Catharanthus roseus (Madagascar periwinkle) was described nearly a decade ago, but the enzyme function remained unknown. We now show by in situ hybridization and immunohistochemistry that the expression in immature leaves is epidermis-specific. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service