Skip to Content
Merck
All Photos(1)

Key Documents

L2145

Sigma-Aldrich

L-Leucine-7-amido-4-methylcoumarin hydrochloride

fluorogenic, ≥98% (TLC), powder

Synonym(s):

H-L-Leu-AMC HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O3 · HCl
CAS Number:
Molecular Weight:
324.80
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

L-Leucine-7-amido-4-methylcoumarin hydrochloride,

Assay

≥98% (TLC)

form

powder

solubility

methanol: 50 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 327 nm; λem 349 nm (pH 8.0)
λex 380 nm; λem 440 nm (Reaction product)

storage temp.

−20°C

SMILES string

Cl.CC(C)C[C@H](N)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C16H20N2O3.ClH/c1-9(2)6-13(17)16(20)18-11-4-5-12-10(3)7-15(19)21-14(12)8-11;/h4-5,7-9,13H,6,17H2,1-3H3,(H,18,20);1H/t13-;/m0./s1

InChI key

VCRXITKKWBOQRZ-ZOWNYOTGSA-N

General description

L-Leucine-7-amido-4-methylcoumarin hydrochloride (Leu-AMC) is a fluorogenic peptidyl substrate for leucine aminopeptidase, an extracellular enzyme found abundantly in natural aquatic systems. This enzyme is one of the various enzymes used by bacteria for protein hydrolysis.

Application

L-Leucine-7-amido-4-methylcoumarin hydrochloride (Leu-AMC) as a substrate analog to determine leucine aminopeptidase activity
L-Leucine-7-amido-4-methylcoumarin hydrochloride has been used:
  • as a substrate in fluorescence-based soil assays
  • to determine leucine aminopeptidase activity of both Plasmodium falciparum M1 (PfA-M1) and PfA-M17 enzymes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzyme activity responses to nutrient loading in subtropical wetlands
Penton CR and Newman S
Biogeochemistry, 84, 83-98 (2007)
Vincent Tardy et al.
Journal of hazardous materials, 411, 125121-125121 (2021-04-17)
Pharmaceutical industry effluents are complex and highly variable in time. Assessing the efficiency of a pharmaceutical industry wastewater treatment plant (WWTP) and the resulting decrease in effluent toxicity and ecological risk is thus not straightforward. We set up an original
Vincent Tardy et al.
Journal of hazardous materials, 411, 125121-125121 (2021-04-17)
Pharmaceutical industry effluents are complex and highly variable in time. Assessing the efficiency of a pharmaceutical industry wastewater treatment plant (WWTP) and the resulting decrease in effluent toxicity and ecological risk is thus not straightforward. We set up an original
C Ryan Penton et al.
Journal of environmental quality, 37(3), 972-976 (2008-05-06)
Enzyme catalyzed reactions are generally considered the rate-limiting step in organic matter degradation and may be significantly influenced by the structure and composition of plant communities. Changes in these rates have the potential to effect long-term peat accumulation and influence
Activity profiling of ectomycorrhiza communities in two forest soils using multiple enzymatic tests
Courty PE, et al.
The New phytologist, 167, 309-319 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service