Skip to Content
Merck
All Photos(5)

Key Documents

M8883

Sigma-Aldrich

4-Methylumbelliferyl phosphate

phosphatase substrate, fluorogenic, ≥98% (HPLC), powder

Synonym(s):

4-Methylumbelliferyl-phosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9O6P
CAS Number:
Molecular Weight:
256.15
Beilstein:
1687229
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Methylumbelliferyl phosphate, phosphatase substrate

description

phosphatase substrate

Assay

≥98% (HPLC)

form

powder

solubility

water: 25 mg/mL, clear to very slightly hazy, colorless

fluorescence

λex 319 nm; λem 384 nm (pH 9.1)
λex 360 nm; λem 449 nm (Reaction product)

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(OP(O)(O)=O)ccc12

InChI

1S/C10H9O6P/c1-6-4-10(11)15-9-5-7(2-3-8(6)9)16-17(12,13)14/h2-5H,1H3,(H2,12,13,14)

InChI key

BCHIXGBGRHLSBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methylumbelliferyl phosphate has been used as a substrate for alkaline phosphatase in the peptide-binding assay. It has also been used as a fluorogenic substrate in enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

4-Methylumbelliferyl phosphate serves as a fluorogenic substrate for calmodulin-dependent protein phosphatase and in kinetic studies of alkaline phosphatase. It is a substrate for alkaline phosphatase in enzyme-linked immunosorbent assay (ELISA) procedures. 4-Methylumbelliferyl phosphate is seven times more sensitive than phenolphthalein monophosphate and 8-13 times more sensitive than with p-nitrophenyl phosphate when used in enzyme immunoassays for the detection of antibodies to human immunodeficiency viruses.

Substrates

Fluorogenic substrate for phosphatases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Bertolini et al.
Journal of pharmaceutical sciences, 96(11), 2931-2944 (2007-08-21)
Oxidative damage to proteins, implicated amongst other in the etiology and progression of Parkinson's disease (PD) and Alzheimer's disease (AD), results in the loss of specific biological protein function. A simple, sensitive, and cost-effective fluorimetric test to assess the antioxidant
N Bouaícha et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 40(11), 1677-1683 (2002-08-15)
Protein phosphatase inhibition assays currently used for the detection of cyanobacterial peptide hepatotoxins in drinking water require an enrichment step using C18 cartridges to achieve lower the detection limit. This paper describes a colorimetric and fluorometric protein phosphatase inhibition method
Laurent Gilardin et al.
Haematologica, 102(11), 1833-1841 (2017-07-29)
Acquired thrombotic thrombocytopenic purpura is a rare and severe disease characterized by auto-antibodies directed against "A Disintegrin And Metalloproteinase with Thrombospondin type 1 repeats, 13
S A Rankin et al.
Journal of dairy science, 93(12), 5538-5551 (2010-11-26)
Standard practices for indirectly assessing the pasteurization status of milk products are primarily based on the thermal inactivation kinetics of the endogenous milk enzyme, alkaline phosphatase (ALP). This assessment provides an invaluable, if not required, tool for both regulatory and
Andréa Toma et al.
Diabetes, 58(2), 394-402 (2008-11-18)
A restricted region of proinsulin located in the B chain and adjacent region of C-peptide has been shown to contain numerous candidate epitopes recognized by CD8(+) T-cells. Our objective is to characterize HLA class I-restricted epitopes located within the preproinsulin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service