257370
7-Amino-4-methylcoumarin
99%
Synonym(s):
Coumarin 120
About This Item
Recommended Products
description
chromophore for enzyme substrates
Assay
99%
form
solid
mp
223-226 °C (lit.)
λmax
354 nm in ethanol
storage temp.
2-8°C
SMILES string
CC1=CC(=O)Oc2cc(N)ccc12
InChI
1S/C10H9NO2/c1-6-4-10(12)13-9-5-7(11)2-3-8(6)9/h2-5H,11H2,1H3
InChI key
GLNDAGDHSLMOKX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service