Skip to Content
Merck
All Photos(1)

Key Documents

J4455

Sigma-Aldrich

JSH-23

≥98% (HPLC), solid

Synonym(s):

4-methyl-N1-(3-phenylpropyl)-1,2-benzenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2
CAS Number:
Molecular Weight:
240.34
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

off-white to gray-pink

solubility

DMSO: >10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N(CCCc2ccccc2)c1c(cc(cc1)C)N

InChI

1S/C16H20N2/c1-13-9-10-16(15(17)12-13)18-11-5-8-14-6-3-2-4-7-14/h2-4,6-7,9-10,12,18H,5,8,11,17H2,1H3

InChI key

YMFNPBSZFWXMAD-UHFFFAOYSA-N

Application

JSH-23 has been used as a nuclear factor κB (NF-κB) p65 inhibitor.

Biochem/physiol Actions

JSH-23 is an inhibitor of NF-kB nuclear translocation.
JSH-23 is an inhibitor of NF-kB nuclear translocation. It inhibits LPS and cytokine-induced nuclear translocation of the p65 subunit of NF-kB as analyzed by EMSA and western blot. The compound displays modest potency (IC50 7.1 uM in RAW 264.7), but has the unique property that it does not affect IkB degradation or recovery. The compound dose dependently inhibits LPS induced expression of cytokines, COX2 and iNOS, and presumably binds to, or interferes with the NLS of p65.

Other Notes

Light and air sensitive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HMGB1 promotes the activation of NLRP3 and caspase-8 inflammasomes via NF-?B pathway in acute glaucoma.
Chi W et al.
Journal of Neuroinflammation, 12, 137-137 (2015)
Macrophage TCF-4 co-activates p65 to potentiate chronic inflammation and insulin resistance in mice.
Kang X et al.
Clinical Science (London, England : 1979), 130, 1257-1257 (2016)
Xia Kang et al.
Clinical science (London, England : 1979), 130(14), 1257-1268 (2016-04-30)
Transcription factor 4 (TCF-4) was recently identified as a candidate gene for the cause of type 2 diabetes, although the mechanisms have not been fully elucidated. In the present study, we demonstrated that the TCF-4 transgene in macrophages aggravated high-fat diet
L Shi et al.
Oncogene, 36(12), 1631-1643 (2016-11-22)
The coordination between cellular differentiation and the mesenchymal/stem transition is essential for both embryo development and neoplasia, suggesting a mechanistic link between these two major processes. In this work we show that miR-127, an embryo-expressing lung miRNA, was prominently induced
Meibao Feng et al.
Molecular cancer, 15(1), 77-77 (2016-12-04)
Lipocalin2 (LCN2) is a secretory protein that is aberrantly expressed in several types of cancer and has been involved in metastatic progression. However, neither mechanisms nor the role that LCN2 plays in the metastasis of colorectal cancer are clear. LCN2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service