Skip to Content
Merck
All Photos(2)

Key Documents

481406

Sigma-Aldrich

NF-κB Activation Inhibitor

The NF-κB Activation Inhibitor, also referenced under CAS 545380-34-5, controls the biological activity of NF-κB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

NF-κB Activation Inhibitor, 6-Amino-4-(4-phenoxyphenylethylamino)quinazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N4O
CAS Number:
Molecular Weight:
356.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(CCc3ccc(cc3)Oc4ccccc4)c1ncnc2c1cc(cc2)N

InChI

1S/C22H20N4O/c23-17-8-11-21-20(14-17)22(26-15-25-21)24-13-12-16-6-9-19(10-7-16)27-18-4-2-1-3-5-18/h1-11,14-15H,12-13,23H2,(H,24,25,26)

InChI key

IBAKVEUZKHOWNG-UHFFFAOYSA-N

General description

A cell-permeable quinazoline compound that acts as a highly potent inhibitor of NF-κB transcriptional activation (IC50 = 11 nM in Jurkat cells) and LPS-induced TNF-α production (IC50 = 7 nM in murine splenocytes). Shown to exhibit anti-inflammatory effect on carrageenin-induced paw edema in rat. A 10 mM (1 mg/281 µl) solution of NF-κB Activation Inhibitor (Cat. No. 481407) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NF-κB transcriptional activation
Product does not compete with ATP.
Reversible: no
Target IC50: 11 nM against NF-κB transcriptional activation in Jurkat cells

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Tobe, M., et al. 2003. Bioorg. Med. Chem.11, 383.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Seongjae Kim et al.
Journal of microbiology and biotechnology, 27(10), 1820-1826 (2017-08-03)
Lipoteichoic acid (LTA), a cell wall component of gram-positive bacteria, is recognized by Toll-like receptor 2, expressed on certain mammalian cell surfaces, initiating signaling cascades that include nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB) and mitogen-activated protein kinase. There
Pascal Baret et al.
BioFactors (Oxford, England), 43(4), 577-592 (2017-05-26)
Diabetes and obesity are strongly associated with increased levels of circulating advanced glycation end products (AGEs) and reactive oxygen species (ROS). These two molecular phenomena affect the physiology of adipose tissue, a biological driver of the metabolic syndrome, leading to
Erawan Borkham-Kamphorst et al.
Liver international : official journal of the International Association for the Study of the Liver, 31(5), 656-665 (2011-04-05)
Lipocalin-2 (LCN2) belongs to the lipocalin superfamily, sharing a barrel-shaped tertiary structure with a hydrophobic pocket and an ability to bind lipophilic molecules. LCN2 has recently emerged as an important modulator of cellular homeostasis in several organs, i.e. heart, lung
Chi-Shuan Fan et al.
Cells, 11(2) (2022-01-22)
M2-polarization and the tumoricidal to tumor-promoting transition are commonly observed with tumor-infiltrating macrophages after interplay with cancer cells or/and other stroma cells. Our previous study indicated that macrophage M2-polarization can be induced by extracellular HSP90α (eHSP90α) secreted from endothelial-to-mesenchymal transition-derived
Veronica De Paolis et al.
International journal of molecular sciences, 23(14) (2022-07-28)
The cellular heterogeneity of the tumor environment of breast cancer (BC) is extremely complex and includes different actors such as neoplastic, stromal, and immunosuppressive cells, which contribute to the chemical and mechanical modification of the environment surrounding the tumor-exasperating immune-escaping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service