Skip to Content
Merck
All Photos(1)

Key Documents

I8779

Sigma-Aldrich

Insulin-like Growth Factor-I from mouse

≥98% (SDS-PAGE & HPLC), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IGF-I

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Insulin-like Growth Factor-I from mouse, IGF-I, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE and HPLC)

form

lyophilized powder

quality

endotoxin tested

mol wt

predicted mol wt ~7.6 kDa

packaging

glass bottle of 50 μg (in glass insert)

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 ng/mg (LAL test)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IGF1(3479)
mouse ... Igf1(16000)

General description

Insulin-like growth factor I (IGF-I) belongs to the insulin-like growth factor (IGF; see MIM 147440) family of signaling molecules. IGF-I is also known as somatomedin. In the developing embryo IGF-I is expressed primarily by mesenchymal-derived cells.

Application

Insulin-like Growth Factor-I from mouse has been used:
  • for the preparation of growth factor cocktail
  • as a media supplement for mouse embryonic fibroblasts culture
  • as a media supplement to serve as a positive control to investigate
  • the effects of Angelica sinensis on myotubes

Biochem/physiol Actions

Insulin-like growth factor I (IGF-I) plays critical roles in cellular energy metabolism and in growth and development, especially prenatal growth.

IGF-I is secreted from the liver into circulation in a process regulated by pituitary growth hormone (GH) and so it mediates the growth-promoting activity of GH. After birth IGF-I expression in most extrahepatic tissues declines and hepatic expression of IGF-I becomes GH-regulated. Expression of IGF-I outside the liver is regulated differently, depending on the specific tissues. For example, gonadotropins and sex steroids regulate IGF-I expression within the reproductive system, while parathyroid hormone and sex steroids regulate IGF-I expression in bone. IGF-I is produced in several human tumors. IGF-I is mitogenic for a variety of cells including fibroblasts, osteoblasts, smooth muscle cells, fetal brain cells, neuroglial cells, and erythroid progenitor cells. IGF-I exerts its actions exclusively through the IGF-I receptor (IGF-IR). IGF-I induces endothelial cell migration and is involved in the regulation of angiogenesis.
Potent mitogenic growth factor that mediates the growth-promoting activities of growth hormone postnatally. Mouse and human IGF-I share 97% sequence identity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activty is measured in a serum-free cell proliferation assay using the human breast carcinoma cell line, MCF-7.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelica Sinensis promotes myotube hypertrophy through the PI3K/Akt/mTOR pathway
Yeh TS, et al.
BMC Complementary and Alternative Medicine, 14(1), 144-144 (2014)
Early tissue patterning recreated by mouse embryonic fibroblasts in a three-dimensional environment
Tissue Engineering: Part A, 15(1), 45-54 (2008)
Simone Frisari et al.
International journal of molecular sciences, 23(3) (2022-02-16)
FOXG1 is an ancient transcription factor gene mastering telencephalic development. A number of distinct structural FOXG1 mutations lead to the "FOXG1 syndrome", a complex and heterogeneous neuropathological entity, for which no cure is presently available. Reconstruction of primary neurodevelopmental/physiological anomalies
Julie Chenal et al.
The European journal of neuroscience, 27(1), 53-65 (2007-12-21)
MCT2 is the main neuronal monocarboxylate transporter essential for facilitating lactate and ketone body utilization as energy substrates. Our study reveals that treatment of cultured cortical neurons with insulin and IGF-1 led to a striking enhancement of MCT2 immunoreactivity in
Soumya Pati et al.
BioMed research international, 2014, 503162-503162 (2014-09-26)
The striatum is considered to be the central processing unit of the basal ganglia in locomotor activity and cognitive function of the brain. IGF-1 could act as a control switch for the long-term proliferation and survival of EGF+bFGF-responsive cultured embryonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service