Skip to Content
Merck
All Photos(1)

Key Documents

I8773

Sigma-Aldrich

Anti-Insulin-Like Growth Factor-I antibody produced in goat

affinity isolated antibody

Synonym(s):

Anti-IGF-I

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: 3-12 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGF1(3479)

Related Categories

General description

Insulin-like growth factors -1 and -2 are important regulators of signalling that mediates growth and metabolism in cells. Insulin-like growth factor -1 shares structural homology with proinsulin and is mainly produced by liver. IGF-1 acts via specific receptor, IGF-1R, a heterodimeric tyrosine kinase receptor that signals to various second messenger pathways such as MAPK, Raf/Ras and PI3K cascades. The effect of IGF-1 is by endocrine, paracrine and autocrine signalling. The effects of IGF-1 signalling are important at many developmental levels from cells to organ to organism, growth, differentiation, anti-apoptosis, proliferation and transformation. Irregular IGF-1 signaling has been implicated in many cancers including melanoma, breast, lung, colon and prostate
Anti-IGF-I recognizes human insulin-like growth factor I. The antibody does not react with human IGF-II.

Specificity

The antibody neutralizes the biological activity of recombinant human IGF-I.

Immunogen

recombinant human IGF-I expressed in Escherichia coli

Application

Anti- IGF-1 antibody may be used for neutralization reactions, the ND50 of the antibody is 3-12 μg/ml. For immunoblotting a working antibody concentration of 0.1-0.2 μg/ml.

Physical form

Lyophilized from a 0.2 μm filtered solution of phosphate buffered saline, pH 7.4, with 5% trehalose.

Preparation Note

Affinity isolated by IGF-I affinity chromatography.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S E Hankinson et al.
Lancet (London, England), 351(9113), 1393-1396 (1998-05-21)
Insulin-like growth factor (IGF)-I, a mitogenic and antiapoptotic peptide, can affect the proliferation of breast epithelial cells, and is thought to have a role in breast cancer. We hypothesised that high circulating IGF-I concentrations would be associated with an increased
Aimei Zhong et al.
Journal of cellular and molecular medicine, 18(7), 1257-1266 (2014-04-12)
True macromastia is a rare but disabling condition characterized by massive breast growth. The aetiology and pathogenic mechanisms for this disorder remain largely unexplored because of the lack of in vivo or in vitro models. Previous studies suggested that regulation
H Yu et al.
Journal of the National Cancer Institute, 91(2), 151-156 (1999-01-29)
Insulin-like growth factors (IGFs), in particular IGF-I and IGF-II, strongly stimulate the proliferation of a variety of cancer cells, including those from lung cancer. To examine the possible causal role of IGFs in lung cancer development, we compared plasma levels
J M Chan et al.
Science (New York, N.Y.), 279(5350), 563-566 (1998-02-07)
Insulin-like growth factor-I (IGF-I) is a mitogen for prostate epithelial cells. To investigate associations between plasma IGF levels and prostate cancer risk, a nested case-control study within the Physicians' Health Study was conducted on prospectively collected plasma from 152 cases
J Dupont et al.
Hormone research, 55 Suppl 2, 22-26 (2001-10-31)
The insulin receptor (IR) and the insulin-like growth factor I receptor (IGF-IR) belong to the same subfamily of receptor tyrosine kinases with two extracellular alpha-subunits and two transmembrane beta-subunits. They share a high similarity of structure and intracellular signalling events.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service