SRP3329
EGF from mouse
Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Synonym(s):
URG, Urogastrone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
mouse
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
potency
≥1x10^7 units/mg
mol wt
6.0 kDa
packaging
pkg of 500 μg
impurities
endotoxin, tested
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... EGF(13645)
General description
Epidermal growth factor (EGF) is a potent growth factor that stimulates the proliferation of various epidermal and epithelial cells. The gene encoding this protein is localized on mouse chromosome 3. Recombinant murine EGF is a 6.0kDa globular protein containing 53 amino acid residues, including three intramolecular disulfide-bonds.
Biochem/physiol Actions
Epidermal growth factor (EGF) has been shown to inhibit gastric secretion, and to be involved in wound healing. EGF signals through a receptor known as c-erbB, which is a class I tyrosine kinase receptor. This receptor also binds with transforming growth factor-α (TGF-α) and VGF (vaccinia virus growth factor). Injection of EGF into neonatal and adult rats has been shown to induce neurobehavioral deficits related to schizophrenia.
Physical form
Lyophilized from 10 mM Sodium Phosphate, pH 8.5.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Epidermal growth factor therapy and wound healing--past, present and future perspectives.
Hardwicke J
The Surgeon : Journal of the Royal Colleges of Surgeons of Edinburgh and Ireland, 6(3), 172-177 (2008)
Reactive oxygen species (ROS) are essential mediators in epidermal growth factor (EGF)-stimulated corneal epithelial cell proliferation, adhesion, migration, and wound healing.
Huo Y
Experimental Eye Research, 89(6), 876-886 (2009)
Gene for lymphoid enhancer-binding factor 1 (LEF1) mapped to human chromosome 4 (q23-q25) and mouse chromosome 3 near Egf.
Milatovich A
Genomics, 11(4), 1040-1048 (1991)
Transforming growth factor alpha and epidermal growth factor in protection and healing of gastric mucosal injury.
Konturek SJ
Scandinavian Journal of Gastroenterology, 27(8), 649-655 (1992)
Wei Tu et al.
Journal of cellular and molecular medicine, 25(11), 5220-5237 (2021-05-06)
MicroRNAs have emerged as essential regulators in the biological process of liver regeneration by modulating the post-transcriptional expression of the target genes. In the present study, we found miR-20a expression is decreased remarkably in three rodent liver regeneration models using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service