Skip to Content
Merck
All Photos(1)

Documents

HPA008069

Sigma-Aldrich

Anti-LAMB3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Kalinin B1 chain antibody produced in rabbit, Anti-Laminin 5 β 3 antibody produced in rabbit, Anti-Laminin B1k chain antibody produced in rabbit, Anti-Laminin subunit β-3 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KLVTSMTKQLGDFWTRMEELRHQARQQGAEAVQAQQLAEGASEQALSAQEGFERIKQKYAELKDRLGQSSMLGEQGARIQSVKTEAEELFGETMEMMDRMKDMELELLRGSQAIMLRSADLTGLEKRVEQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMB3(3914)

Immunogen

Laminin subunit β-3 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-LAMB3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

LAMB3 (Laminin subunit beta-3) gene encodes the β3-subunit that forms laminin-5 along with α and γ subunits. It belongs to the family of basement membrane proteins and is implicated in several cellular processes, such as cell differentiation, migration, adhesion, proliferation and survival. It functions as an oncogene in cervical squamous cell carcinoma. Mutations in this gene have been linked to generalized atrophic benign epidermolysis bullosa

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71223

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noriko Yamamoto et al.
International journal of oncology, 42(5), 1523-1532 (2013-03-14)
Cervical cancer is one of the most common cancers in women. More than 275,100 women die from cervical cancer each year. Cervical squamous cell carcinoma (cervical SCC), one of the most frequent types of cervical cancers, is associated with high-risk
Takashi Kinoshita et al.
Oncotarget, 3(11), 1386-1400 (2012-11-20)
Recent our microRNA (miRNA) expression signature revealed that expression of microRNA-218 (miR-218) was reduced in cancer tissues, suggesting a candidate of tumor suppressor in head and neck squamous cell carcinoma (HNSCC). The aim of this study was to investigate the
J E Mellerio et al.
The British journal of dermatology, 139(2), 325-331 (1998-10-10)
Pathogenetic mutations in the genes encoding the hemidesmosome-anchoring filament complex proteins, laminin-5 and the 180 kDa bullous pemphigoid antigen, have been identified in patients with the inherited mechanobullous disease, junctional epidermolysis bullosa (EB). Furthermore, there is some evidence to suggest
J A McGrath et al.
The Journal of investigative dermatology, 104(4), 467-474 (1995-04-01)
The anchoring filament component laminin 5 (kalinin/nicein) is a candidate protein for mutations in some hereditary blistering skin disorders. In this study, laminin 5 expression was assessed in a family with generalized atrophic benign epidermolysis bullosa, a non-lethal variant of
Sidharth V Puram et al.
Cell, 171(7), 1611-1624 (2017-12-05)
The diverse malignant, stromal, and immune cells in tumors affect growth, metastasis, and response to therapy. We profiled transcriptomes of ∼6,000 single cells from 18 head and neck squamous cell carcinoma (HNSCC) patients, including five matched pairs of primary tumors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service