Skip to Content
Merck
All Photos(1)

Key Documents

CAX2-S0000

Millipore

CellASIC ONIX2 Microfluidic System

Synonym(s):

CellASIC ONIX2, Microfluidic System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NC.51
Pricing and availability is not currently available.

manufacturer/tradename

CellASIC® ONIX

W × D × H

330 mm × 306 mm × 108 mm

shipped in

ambient

General description

CellASIC® ONIX2 Microfluidic System is an automated platform for precise manipulation of multiple key cell culture parameters, enabling measurement of cellular responses to pre-programmed media, temperature, and gas environment changes. It provides detailed information about live cell processes with ultra-high quality images and complete environment control. The system utilizes high quality, optically clear microfluidic plates and intuitive software, while integrating with a broad range of inverted microscopes to allow continuous, high magnification observation of live cells—as they react to their environment in time.

Includes:
  • Controller System: Small footprint, integrated microincubator controller maintains fluid movement, reagent additions, temperature, and gas conditions.
  • Microfluidic Plates: Application specific plates bring new cell culture capabilities for live cell imaging.
  • Manifold: Low-profile, high quality, high optical clarity culture chambers and manifold, mount easily on an inverted microscope—without the need for a bulky environmental chamber.
  • Software: Intuitive software enables quick and easy set-up of detailed protocols for truly automated hands-free cell culture.

Controller System:
  • Automatically control flow rates, gas and temperature shifts, standing gradients, nutrient/drug additions, and media changes.
  • Eight critical cell culture parameters can be controlled by the system.
  • Temperature, oxygen level, growth factors, humidity, pH, osmolality, glucose, ECM and adhesion.

Advantages of the CellASIC® ONIX2 microfluidic plates:
  • High resolution imaging through optical glass bottom and chamber structures.
  • Ability to run multiple independent experiments simultaneously.
  • Continuous access to nutrients and ongoing removal of waste, promoting optimal cell health.

Manifold Advantages:
  • Manifold seals to the microfluidic plate via “one-touch” vacuum mechanism.
  • Pressure-driven flow of liquids to the cell chamber provides high precision even at very small volumes.
  • No contact between the flow system and the biological solutions (on the plate) prevents contamination.
  • Low-profile manifold is easy to adapt to any inverted microscope stage.

The software offers:
  • Clearly defined protocols with all experimental parameters saved in one file.
  • Convenience and reproducibility.
  • Consistency and reliability.
  • Error checking, self test, and data logging all make for better troubleshooting and serviceability.

Application

  • Yeast single cell response.
  • Bacterial single cell response.
  • Bacterial biofilm dynamics.
  • Cell response to hypoxic environments.
  • Cell response to changing media conditions, drugs, and other stimulants.
  • Host-pathogen interactions.
  • Chemotaxis/migration in response to chemogradient.
  • Setting optimal culture parameters for difficult cell types or long term experiments.
  • Microscopy of 3D cell culture.

Legal Information

CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is the Premixed Gas Regulator Accessory for the ONIX2 (CAX2-ABR00) compatible with the older CellASIC ONIX model for gas control? Additionally, can the ONIX1 be fitted with a separate gas line and regulator, similar to the ONIX2?

    1 answer
    1. The Premixed Gas Regulator Accessory for the ONIX2 (CAX2-ABR00) is not compatible with the older CellASIC ONIX model for gas control. However, the ONIX1 can be fitted with a separate gas line and regulator, similar to the ONIX2. The original gas connection method for the ONIX1 is no longer available, but it can be retrofitted with a separate gas line and regulator. Additionally, the manual recommends using the Colder Products Co. 4 mm valved coupling insert part no. PMCD20025 for fitting to connect low gas-permeable tubing to the controller.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service