MDCI10000
Millicell® Digital Cell Imager
Synonym(s):
Cell Imager, Millicell DCI, cell counter, hemacytometer, hemocytometer
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Recommended Products
L × W × H
6 in. × 11 in. × 12 in., Millicell® Digital Cell Imager
weight
5.67 lbGross weight
compatibility
USB/Ethernet/Wi-Fi
General description
Assess your cell cultures with ease. Our Millicell® DCI instrument enables more efficient execution of the repetitive daily techniques associated with cell passaging. Quickly and objectively assess confluency, morphology, and growth trends for more consistent cell cultures.
Remove subjectivity by using accurate confluency measurements!
WiFi compatible with included EDIMAX extneral USB, Ethernet compatible.
When purchasing Our Millicell® DCI instrument please also choose a software subscription as well.
Remove subjectivity by using accurate confluency measurements!
WiFi compatible with included EDIMAX extneral USB, Ethernet compatible.
When purchasing Our Millicell® DCI instrument please also choose a software subscription as well.
- Free Trial Software Subscription: MDCI1TRIAL
- 1 Year Software Subscription: MDCI1T1YR
- Lifetime Software Subscription: MDCI1T1LIF
Features and Benefits
- Faster analysis
- Reduced user bias
- Confluency, cell count, and morphology
- Hemocytometer or in-vessel measurement
- Two-click image capture
- Convenient, web-based cloud service for data storage and archiving
Components
- Instrument
- Power supply. Replacement available: MDCI1USBD0N
Legal Information
Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service