Skip to Content
Merck
All Photos(1)

Key Documents

GERPN800E

Amersham ECL Rainbow Marker - Full range

Full Range, Cytiva RPN800E

Synonym(s):

molecular weight markers, protein markers, protein ladders and standards for SDS-PAGE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331
NACRES:
NA.25
Pricing and availability is not currently available.

form

liquid

feature

wetted part: no
Full Range

packaging

pkg of 250 μL

manufacturer/tradename

Cytiva RPN800E

color

Brownish-red

solubility

cold water: soluble (Easily soluble in cold and hot water)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Rainbow Molecular Weight Markers have been enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. The ready-to-load markers provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination. The bright, distinctive colors of the markers allow easier confirmation of transfer to blotting membranes and orientation.

Rainbow Molecular Weight Markers comprise a mixture of individually colored proteins that are combined to produce bands of equal color intensity. The approximate protein concentration is 1 to 2 mg/mL, depending on the product. Each 250 μl pack size is sufficient for use with 50 minigels (10 x 8 cm) or 25 large gels (20 x 20 cm).

Application

For industrial applications: Analytical reagent, research
Amersham ECL Rainbow Marker - Full range has been used as a molecular weight marker for immunoblotting and sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE).[1][2][3]

Features and Benefits

  • Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins.without staining
  • High intensity, prestained multicolored bands allow distinction of closely related proteins.
  • Full-Range Rainbow Markers (Mr 12 000 to 225 000) use ten separate proteins with six different colors.
  • High-Range Rainbow Markers (Mr 12 000 to 225 000) use eight separate proteins with six different colors.
  • Low-Range Rainbow Markers (Mr 3 500 to 38 000) use seven separate proteins with five different colors.
  • For use on SDS-PAGE

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva
Rainbow is a trademark of Cytiva

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonia production from amino acid-based biomass-like sources by engineered Escherichia coli
Yosuke M, et al.
AMB Express (2017)
Mesenchymal stem cells/multipotent stromal cells (MSCs) are glycolytic and thus glucose is a limiting factor of in vitro models of MSC starvation
Austin N, et al.
Stem Cell Research & Therapy (2016)
Linear immunoglobulin A bullous dermatosis developing during late pregnancy
Naoyuki I, et al.
The Journal of Dermatology, 44(3) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service