GERPN756E
Amersham™ ECL™ Rainbow™ Marker - Full range
High Range, Cytiva RPN756E
Synonym(s):
ECL Rainbow Marker
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
feature
wetted part: no
High Range
packaging
pkg of 250 μL
manufacturer/tradename
Cytiva RPN756E
color
Brownish-red
solubility
cold water: soluble (Easily soluble in cold and hot water)
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Rainbow™ Molecular Weight Markers have been enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. The ready-to-load markers provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination. The bright, distinctive colors of the markers allow easier confirmation of transfer to blotting membranes and orientation.
Rainbow™ Molecular Weight Markers comprise a mixture of individually colored proteins that are combined to produce bands of equal color intensity. The approximate protein concentration is 1 to 2 mg/mL, depending on the product. Each 250 μl pack size is sufficient for use with 50 minigels (10 x 8 cm) or 25 large gels (20 x 20 cm).
Rainbow™ Molecular Weight Markers comprise a mixture of individually colored proteins that are combined to produce bands of equal color intensity. The approximate protein concentration is 1 to 2 mg/mL, depending on the product. Each 250 μl pack size is sufficient for use with 50 minigels (10 x 8 cm) or 25 large gels (20 x 20 cm).
Application
For industrial applications: Analytical reagent, research
Features and Benefits
- Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins.without staining
- High intensity, prestained multicolored bands allow distinction of closely related proteins.
- Full-Range Rainbow™ Markers (Mr 12 000 to 225 000) use ten separate proteins with six different colors.
- High-Range Rainbow™ Markers (Mr 12 000 to 225 000) use eight separate proteins with six different colors.
- Low-Range Rainbow™ Markers (Mr 3 500 to 38 000) use seven separate proteins with five different colors.
- For use on SDS-PAGE
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva
Rainbow is a trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service