Skip to Content
Merck
All Photos(1)

Documents

T7951

Sigma-Aldrich

Tau Protein Ladder, 6 isoforms human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Tau Protein Ladder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

36800
39700
40000
42600
42900
45900

composition

dodecyl sulphate sodium salt, 1-5%
glycerine, 20-30%
mercaptoethanol, 10-20%

technique(s)

immunoelectrophoresis: 10-20 μL using recombinant Tau protein marker
western blot: 2-5 μL using recombinant Tau protein marker

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MAPT(4137)

General description

Research area: Neuroscience

Tau gene, spanning 100 kb with 16 exons, is mapped to human chromosome 17q21. The six alternative splice variants of protein ranging in size from 352-441 amino acids have been identified in human adult brain. Tau is a member of the microtubule-associated protein (MAP) family. It is predominantly expressed in neurons.

Application

Tau Protein Ladder, 6 isoforms human has been used as a sample in immunomagnetic reduction (IMR) assay. It has also been used as positive control in western blotting.

Biochem/physiol Actions

In neurons, tau protein plays an important role in maintaining microtubule assembly and stability. It acts as a connector between microtubules and other cytoskeletal elements or proteins. Overexpression of the protein has been observed in Alzheimer′s disease and various neurodegenerative disorders denoted as ‘tauopathies′. Thus, pathological tau proteins act as a potential biomarker in the neurodegenerative process.

Packaging

A 50 μL vial of Tau Protein Ladder contains 0.25 μg of each of the six isoforms.

Specifications

Contains 6 recombinant Tau proteins expressed in E. coli, which comprise six Tau isoforms with molecular weights of 45,900, 42,600, 42,900, 39,700, 40,000, and 36,800 respectively. No histidine-tags are present in the proteins.

Physical form

Solution in 125 mM Tris-HCl, pH 6.8, containing 4% SDS, 10% 2-mercaptoethanol, 20% glycerol, and 0.004% bromphenol blue.

Storage and Stability

Tightly closed. Keep in a well-ventilated place. Keep locked up or in an area accessible only
to qualified or authorized persons

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Liver,Heart

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nora Lemke et al.
The Journal of biological chemistry, 295(52), 18508-18523 (2020-11-01)
Synapse loss is associated with motor and cognitive decline in multiple neurodegenerative disorders, and the cellular redistribution of tau is related to synaptic impairment in tauopathies, such as Alzheimer's disease and frontotemporal dementia. Here, we examined the cellular distribution of
Tau protein in cerebrospinal fluid: a biochemical marker for axonal degeneration in Alzheimer disease?
Blennow K, et al.
Molecular and Chemical Neuropathology, 26(3), 231-245 (1995)
Rebecca L Mueller et al.
Frontiers in molecular neuroscience, 14, 647054-647054 (2021-04-06)
Over four decades ago, in vitro experiments showed that tau protein interacts with and stabilizes microtubules in a phosphorylation-dependent manner. This observation fueled the widespread hypotheses that these properties extend to living neurons and that reduced stability of microtubules represents
Antibody-mediated clearance of tau in primary mouse microglial cultures requires Fc gamma-receptor binding and functional lysosomes
Andersson C, et al.
Scientific reports, 9(1), 4658-4658 (2019)
Emma S Davies et al.
Cellular and molecular life sciences : CMLS, 79(11), 560-560 (2022-10-22)
Alzheimer's disease is a chronic neurodegenerative disease that accounts for up to 80% of all dementias. Characterised by deteriorations of memory and cognitive function, the key neuropathological features are accumulations of β-amyloid and hyperphosphorylated tau, as 'plaques' and 'tangles', respectively.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service