Skip to Content
Merck
All Photos(1)

Key Documents

S8445

Sigma-Aldrich

SigmaMarker

wide range, mol wt 6,500-200,000 Da

Synonym(s):

protein markers, protein standards

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105337
NACRES:
NA.25

Quality Level

form

lyophilized powder

usage

sufficient for 10-20 large gels (16 × 18 cm)
sufficient for 20-30 mini-gels (10 × 10 cm)
 vial sufficient for 50-100 PhastGels

mol wt

6,500-200,000 Da

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

SigmaMarker has been used as a protein marker in sodium dodecyl sulfate–polyacrylamide gel electrophoresis (SDS-PAGE).
SigmaMarkers are designed for use with Laemmli SDS-PAGE, the PhastSystem (Pharmacia) and Schagger and von Jagow SDS-PAGE (Tricine). SigmaMarkers cover the range of molecular weights common to most proteins or their subunits. The wide, high and low range markers are lyophilized with sample buffer so that they are ready-to-use after reconstitution with deionized water. The markers are formulated to yield a distribution of well-defined bands of approximately equal intensity after electrophoresis and Coomassie blue staining.

Other Notes

Protein (mol. wt.)
Aprotinin, bovine lung (6,500)
α-Lactalbumin, bovine milk (14,200)
Trypsin inhibitor, soybean (20,000)
Trypsinogen, bovine pancreas (24,000)
Carbonic anhydrase, bovine erythrocytes (29,000)
Glyceraldehyde-3-phosphate dehydrogenase, rabbit muscle (36,000)
Ovalbumin, chicken egg (45,000)
Glutamic dehydrogenase, bovine liver (55,000)
Albumin, bovine serum (66,000)
Phosphorylase B, rabbit muscle (97,000)
β-Galactosidase, E. coli (116,000)
Myosin, porcine heart (200,000)
Will yield 12 protein bands with Coomassie Blue staining.

Reconstitution

Reconstitution with 100 μl of deionized water results in a solution containing 2-3.5 mg of protein per ml of 62 mM Tris-HCl, pH 6.8, 1 mM EDTA, 4% sucrose, 10 mM TCEP, 2% SDS, and 0.01% bromophenol blue.

Legal Information

SigmaMarker is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pavel Mokrejs et al.
Journal of cosmetic dermatology, 16(4), e21-e27 (2017-02-07)
Although keratin hydrolysates have become established as standard components in hair and nail cosmetics, studies on the moisturizing effects of keratin hydrolysates do not appear among contemporary literature. To test if adding keratin hydrolysate into an ointment base increases hydration
Sule Keskin et al.
Journal of economic entomology, 111(3), 1454-1460 (2018-03-13)
The objective of this study was to investigate the effects of storage and granary weevil, Sitophilus granarius (L.; Coleoptera: Curculionidae), infestation on pH, protein solubility (PS) and gel electrophoresis properties of meal and roller-milled flours of hard (Ceyhan-99 cv.) and
François Potin et al.
Journal of food science, 84(12), 3682-3690 (2019-11-07)
The purpose of this paper was to study the extraction conditions of hemp proteins from undelipidated press-cakes. The effects of different hydration conditions on protein recovery yield and polypeptide profile were evaluated: pH (2 to 12), ionic strength (0 to
Thermal denaturation of pea globulins (Pisum sativum L.)-molecular interactions leading to heat-induced protein aggregation.
Mession JL, et al.
Journal of Agricultural and Food Chemistry, 61, 1196-1204 (2013)
Natalia Bourguignon et al.
Ecotoxicology and environmental safety, 175, 19-28 (2019-03-18)
In the present study the polycyclic aromatic hydrocarbon removal and metabolic adaptation of Amycolatopsis tucumanensis DSM 45259 were investigated. Analysis of one-dimensional gel electrophoresis of crude cell extracts revealed differential synthesis of proteins which were identified by MALDI-TOF. To elucidate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service