GE17-1408-01
HiTrap® Desalting Columns
Cytiva 17-1408-01, pack of 5 × 5 mL
Synonym(s):
Purification Columns
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151816
NACRES:
NA.56
Recommended Products
packaging
pack of 5 × 5 mL
manufacturer/tradename
Cytiva 17-1408-01
storage condition
dry media
parameter
42 psi
bed size
16 mm × 25 mm
bed volume
5 mL
column I.D.
16 mm
matrix
cross-linked dextran
particle size
>20 μm (dry)
cleaning
2-13
working range
2-13
suitability
suitable for bioprocess medium
General description
HiTrap® Desalting is a ready-to-use 5 mL column, prepacked with Sephadex® G-25 Superfine, for fast and convenient desalting and buffer exchange.
HiTrap® Desalting is a prepacked, ready to use column for group separation between high and low molecular weight substances, i.e., buffer exchange prior to or after different chromatographic steps, removal of low molecular weight contaminants or removal of reagents to terminate a reaction.
HiTrap® Desalting is a prepacked, ready to use column for group separation between high and low molecular weight substances, i.e., buffer exchange prior to or after different chromatographic steps, removal of low molecular weight contaminants or removal of reagents to terminate a reaction.
Application
Desalting and buffer exchange with Sephadex® G-25.
Features and Benefits
- Handles sample volumes of 1.5 mL, or larger volumes by connecting up to five columns in series.
- Replaces dialysis as the method of choice for desalting.
- Desalting requires less than one minute due to the high rigidity of Sephadex® G-25 and the optimized packing technology.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (4 to 8, Used Media 20% Ehtanol or 0.01 M NaOH)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Other Notes
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.
Legal Information
HiTrap is a registered trademark of Cytiva
Sephadex is a registered trademark of Cytiva
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service