GE17-0020-01
Sephadex® G-15
Cytiva 17-0020-01, pack of 100 g
Synonym(s):
Sephadex gel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
autoclavable In wet form (pH 7.0) at 120°C for 30 min
packaging
pack of 100 g
manufacturer/tradename
Cytiva 17-0020-01
parameter
flow rate (The beads behave as rigid spheres obeying Darcy′s Law.)
matrix
cross-linked dextran
particle size
40-120 μm (dry)
60-180 μm (wet)
working range
2-13
General description
Sephadex® G-15 is well established gel filtration medium for desalting and buffer exchange of peptides and small biomolecules > 1500 molecular weight.
Application
Sephadex® is a gel filtration medium prepared by crosslinking Dextran with epichlorohydrin. Different types of Sephadex® differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex® G-15 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex® G-50 is available in 4 different particle sizes (Course, Medium, Fine & Superfine) and Superfine has the smallest bead size for higher efficiency with shorter diffusion distances. Cource and Medium are are preferred for large scale group separations at where high flow rates and Low operating pressures are required.
Features and Benefits
- Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
- Suitable for buffer exchange and cleanup of biological samples, for example peptides, small Proteins, and oligosaccharides larger than 1 500 molecular weight
- Efficient removal of contaminants such as salts, dyes, and radioactive labels
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sephadex is a registered trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service