Skip to Content
Merck
All Photos(1)

Key Documents

RIN-m

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 95071701

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

rat pancreas

packaging

tube of 5 μg 95071701-DNA-5UG
pkg of vial of cells 95071701-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Somatostatin, insulin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat islet cell tumour

Cell Line Description

Established from a transplantable pancreatic tumour of the islets of Langerhans. The tumour originated in an irradiated NEDH inhibited albino rat and was maintained by serial transplantation in Nedh rats. After transplantation into athymic nude mice the cell line RIN-m was established from the fourth nude mouse heterotransplant, 9 days after the tumour was removed cells were exposed to cystine-free medium resulting in the complete elimination of fibroblast-like cells. RIN-m cells secrete insulin and somatostatin. High levels of L-dopa-decarboxylase and membrane bound secretory granules were detected. RIN-5F (95090402) was established as a secondary clone from RIN-m.

Application

Study of biology of pancreatic islet cells, synthesis, storage and secretion of insulin and somatostatin

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service